Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423929_g1
          See other ZFP335 GE Assays ›
          Gene Symbol:
          Zfp335
          Gene Name:
          zinc finger protein 335
          Gene Aliases:
          Nif, Nif1, Znf335
          Chromosome Location:
          Chr.3: 161357201 - 161378075 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423929_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp335
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008762559.1 XP_008760781.1 25 - 26 4696 56
          XM_008762560.1 XP_008760782.1 26 - 27 5370 56
          XM_008762561.1 XP_008760783.1 25 - 26 4693 56
          XM_008762563.1 XP_008760785.1 26 - 27 4570 56
          XM_008762564.1 XP_008760786.1 26 - 27 4567 56

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of neuroblast proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          brain development
          regulation of gene expression
          cerebral cortex neuron differentiation
          regulation of gene expression, epigenetic
          neuron projection morphogenesis
          brain morphogenesis
          positive regulation of lymphocyte proliferation
          regulation of neurogenesis
          positive regulation of neurogenesis
          regulation of histone H3-K4 methylation
          histone H3-K4 trimethylation
          transcription cis-regulatory region binding
          RNA polymerase II core promoter sequence-specific DNA binding
          nucleic acid binding
          protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P412121 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412121 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412121 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P412121 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P412121 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851701 - -
          Affymetrix Rat Gene 1.0 ST Array 10851704 - -
          Affymetrix Rat Gene 1.0 ST Array 10851698 - -
          Affymetrix Rat Gene 1.0 ST Array 10851694 - -
          Affymetrix Rat Gene 1.0 ST Array 10851690 - -
          Affymetrix Rat Gene 1.0 ST Array 10851703 - -
          Affymetrix Rat Gene 1.0 ST Array 10851707 - -
          Affymetrix Rat Gene 1.0 ST Array 10851693 - -
          Affymetrix Rat Gene 1.0 ST Array 10851700 - -
          Affymetrix Rat Gene 1.0 ST Array 10851695 - -
          Affymetrix Rat Gene 1.0 ST Array 10851696 - -
          Affymetrix Rat Gene 1.0 ST Array 10851699 - -
          Affymetrix Rat Gene 1.0 ST Array 10851697 - -
          Affymetrix Rat Gene 1.0 ST Array 10851705 - -
          Affymetrix Rat Gene 1.0 ST Array 10851691 - -
          Affymetrix Rat Gene 1.0 ST Array 10851702 - -
          Affymetrix Rat Gene 1.0 ST Array 10851706 - -
          Affymetrix Rat Gene 1.1 ST Array 10851689 - -
          Rat Genome 230 2.0 Array 1375013_PM_at - -
          Rat Genome 230 2.0 Array 1375013_at - -
          Rat Genome 230 2.0 Array 1381609_at - -
          Rat Genome 230 2.0 Array 1381609_PM_at - -
          Affymetrix HT Rat Focus Array 1381609_PM_at - -
          Affymetrix HT Rat Focus Array 1381609_at - -
          Rat Expression Set 230 1381609_at - -
          Rat Expression Set 230 1381609_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851689 - -
          Rat Genome U34 Set rc_AA996814_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375216 - -
          RatRef-12 Expression BeadChip ILMN_58745 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0