Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01425969_m1
          See other LOC103690006 GE Assays ›
          Gene Symbol:
          LOC103690006 Dusp11
          Gene Name:
          RNA/RNP complex-1-interacting phosphatase
          dual specificity phosphatase 11 (RNA/RNP complex 1-interacting)
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.4: 117797735 - 117814737 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 117617640 - 117631674 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01425969_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103690006
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763101.1 XP_008761323.1 1 - 2 235 65
          Gene Symbol: Dusp11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025650.1 NP_001020821.1 1 - 2 156 65
          GenBank mRNA BC098712.1 - 1 - 2 156 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003235.rn.1, TC0400003241.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC103690006
          Click here to find siRNAs for Gene Dusp11

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA metabolic process
          peptidyl-tyrosine dephosphorylation
          polynucleotide 5' dephosphorylation
          RNA binding
          polynucleotide 5'-phosphatase activity
          protein tyrosine phosphatase activity
          protein tyrosine/serine/threonine phosphatase activity
          phosphatase activity
          nucleotide phosphatase activity, acting on free nucleotides

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21892 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5815 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109677 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109677 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1003072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17915 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863708 - -
          Affymetrix Rat Gene 1.0 ST Array 10863700 - -
          Affymetrix Rat Gene 1.0 ST Array 10863705 - -
          Affymetrix Rat Gene 1.0 ST Array 10863702 - -
          Affymetrix Rat Gene 1.0 ST Array 10863701 - -
          Affymetrix Rat Gene 1.0 ST Array 10863706 - -
          Affymetrix Rat Gene 1.0 ST Array 10863704 - -
          Affymetrix Rat Gene 1.0 ST Array 10863699 - -
          Affymetrix Rat Gene 1.0 ST Array 10863709 - -
          Affymetrix Rat Gene 1.0 ST Array 10863703 - -
          Affymetrix Rat Gene 1.0 ST Array 10863707 - -
          Affymetrix Rat Gene 1.1 ST Array 10863698 - -
          Rat Genome 230 2.0 Array 1376576_at - -
          Rat Genome 230 2.0 Array 1376576_PM_at - -
          Affymetrix HT Rat Focus Array 1376576_PM_at - -
          Affymetrix HT Rat Focus Array 1376576_at - -
          Affymetrix Rat Clariom S Assay PSR0400023066.rn.1 TC0400003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400023024.rn.1 TC0400003235.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400023068.rn.1 TC0400003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400023023.rn.1 TC0400003235.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400023020.rn.1 TC0400003235.rn.1 RTA TC
          Rat Expression Set 230 1376576_PM_at - -
          Rat Expression Set 230 1376576_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863698 - -
          Rat Genome U34 Set rc_AI232489_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371020 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0