Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426273_m1
          See other CRELD1 GE Assays ›
          Gene Symbol:
          Creld1
          Gene Name:
          cysteine-rich with EGF-like domains 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 145439964 - 145449894 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426273_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Creld1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024783.1 NP_001019954.1 7 - 8 900 72
          XM_006237027.2 XP_006237089.1 6 - 7 1387 72
          XM_006237028.2 XP_006237090.1 6 - 7 945 72
          GenBank mRNA BC097951.1 - 7 - 8 900 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001576.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Creld1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P827677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P827677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P827677 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P827677 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P827677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37970 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857834 - -
          Affymetrix Rat Gene 1.0 ST Array 10857836 - -
          Affymetrix Rat Gene 1.0 ST Array 10857829 - -
          Affymetrix Rat Gene 1.0 ST Array 10857837 - -
          Affymetrix Rat Gene 1.0 ST Array 10857831 - -
          Affymetrix Rat Gene 1.0 ST Array 10857827 - -
          Affymetrix Rat Gene 1.0 ST Array 10857832 - -
          Affymetrix Rat Gene 1.0 ST Array 10857833 - -
          Affymetrix Rat Gene 1.0 ST Array 10857830 - -
          Affymetrix Rat Gene 1.0 ST Array 10857828 - -
          Affymetrix Rat Gene 1.0 ST Array 10857835 - -
          Affymetrix Rat Gene 1.1 ST Array 10857826 - -
          Rat Genome 230 2.0 Array 1388594_at - -
          Rat Genome 230 2.0 Array 1388594_PM_at - -
          Affymetrix HT Rat Focus Array 1388594_at - -
          Affymetrix HT Rat Focus Array 1388594_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011182.rn.1 TC0400001576.rn.1 RTA TC
          Rat Expression Set 230 1388594_at - -
          Rat Expression Set 230 1388594_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857826 - -
          Rat Genome U34 Set rc_AI175044_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22368511 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0