Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426286_m1
          See other ARPC4 GE Assays ›
          Gene Symbol:
          Arpc4
          Gene Name:
          actin related protein 2/3 complex, subunit 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 145330457 - 145340985 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426286_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arpc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106615.2 NP_001100085.2 5 - 6 529 64
          GenBank mRNA BC167049.1 - 5 - 6 517 64
          CK478610.1 - 5 - 6 529 64
          FQ228740.1 - 5 - 6 468 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001569.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament polymerization
          Arp2/3 complex-mediated actin nucleation
          structural constituent of cytoskeleton
          enzyme binding
          protein-macromolecule adaptor activity
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P541456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120752 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60807 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P541456 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P541456 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P541456 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P541456 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47970 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33593 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857763 - -
          Affymetrix Rat Gene 1.0 ST Array 10857765 - -
          Affymetrix Rat Gene 1.0 ST Array 10857764 - -
          Affymetrix Rat Gene 1.0 ST Array 10857766 - -
          Affymetrix Rat Gene 1.1 ST Array 10857762 - -
          Rat Genome 230 2.0 Array 1372577_at - -
          Rat Genome 230 2.0 Array 1372577_PM_at - -
          Affymetrix HT Rat Focus Array 1372577_at - -
          Affymetrix HT Rat Focus Array 1372577_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011059.rn.1 TC0400001569.rn.1 RTA TC
          Rat Expression Set 230 1372577_PM_at - -
          Rat Expression Set 230 1372577_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857762 - -
          Rat Genome U34 Set rc_AA892332_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356401 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0