Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426391_m1
          See other VPS4B GE Assays ›
          Gene Symbol:
          Vps4b
          Gene Name:
          vacuolar protein sorting 4 homolog B (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 26825261 - 26850460 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426391_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vps4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025716.1 NP_001020887.1 8 - 9 1065 66
          GenBank mRNA BC099128.1 - 8 - 9 1065 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001211.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vps4b

          Gene Ontology Categories:

          Function(s) Process(es)

          septum digestion after cytokinesis
          potassium ion transport
          nucleus organization
          endosome organization
          vacuole organization
          mitotic metaphase plate congression
          biological_process
          regulation of centrosome duplication
          positive regulation of G2/M transition of mitotic cell cycle
          endosomal transport
          cholesterol transport
          cytoplasmic microtubule organization
          intracellular cholesterol transport
          endosome to lysosome transport via multivesicular body sorting pathway
          response to lipid
          viral budding via host ESCRT complex
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          regulation of viral process
          protein depolymerization
          negative regulation of cell death
          ubiquitin-independent protein catabolic process via the multivesicular body sorting pathway
          regulation of mitotic spindle assembly
          positive regulation of viral release from host cell
          negative regulation of exosomal secretion
          positive regulation of exosomal secretion
          positive regulation of centriole elongation
          molecular_function
          ATP binding
          protein C-terminus binding
          microtubule-severing ATPase activity
          ATP hydrolysis activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P299671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111455 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299671 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299671 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P299671 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17933 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299671 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35492 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766977 - -
          Affymetrix Rat Gene 1.0 ST Array 10766972 - -
          Affymetrix Rat Gene 1.0 ST Array 10766979 - -
          Affymetrix Rat Gene 1.0 ST Array 10766973 - -
          Affymetrix Rat Gene 1.0 ST Array 10766981 - -
          Affymetrix Rat Gene 1.0 ST Array 10766974 - -
          Affymetrix Rat Gene 1.0 ST Array 10766971 - -
          Affymetrix Rat Gene 1.0 ST Array 10766980 - -
          Affymetrix Rat Gene 1.0 ST Array 10766978 - -
          Affymetrix Rat Gene 1.0 ST Array 10766975 - -
          Affymetrix Rat Gene 1.0 ST Array 10766976 - -
          Affymetrix Rat Gene 1.1 ST Array 10766970 - -
          Rat Genome 230 2.0 Array 1372834_at - -
          Rat Genome 230 2.0 Array 1372834_PM_at - -
          Affymetrix HT Rat Focus Array 1372834_at - -
          Affymetrix HT Rat Focus Array 1372834_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300008096.rn.1 TC1300001211.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008097.rn.1 TC1300001211.rn.1 RTA TC
          Rat Expression Set 230 1372834_PM_at - -
          Rat Expression Set 230 1372834_at - -
          Affymetrix Rat Gene 1.0 ST Array 10766970 - -
          Rat Genome U34 Set AA848378_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21162579 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366691 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0