Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01427493_m1
          See other D2HGDH GE Assays ›
          Gene Symbol:
          D2hgdh
          Gene Name:
          D-2-hydroxyglutarate dehydrogenase
          Gene Aliases:
          RGD1307976
          Chromosome Location:
          Chr.9: 100956569 - 100974393 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01427493_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D2hgdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106926.1 NP_001100396.1 9 - 10 1701 68
          XM_006245520.2 XP_006245582.1 9 - 10 1617 68
          XM_006245521.2 XP_006245583.1 9 - 10 1475 68
          XM_006245522.2 XP_006245584.1 9 - 10 1470 68
          XM_008767365.1 XP_008765587.1 9 - 10 1732 68
          GenBank mRNA BC166559.1 - 8 - 9 1374 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001190.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to manganese ion
          response to zinc ion
          response to cobalt ion
          response to magnesium ion
          cellular protein metabolic process
          response to calcium ion
          UDP-N-acetylmuramate dehydrogenase activity
          flavin adenine dinucleotide binding
          (R)-2-hydroxyglutarate dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P504274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P504274 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P504274 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P504274 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P504274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56136 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925670 - -
          Affymetrix Rat Gene 1.0 ST Array 10925675 - -
          Affymetrix Rat Gene 1.0 ST Array 10925674 - -
          Affymetrix Rat Gene 1.0 ST Array 10925677 - -
          Affymetrix Rat Gene 1.0 ST Array 10925673 - -
          Affymetrix Rat Gene 1.0 ST Array 10925676 - -
          Affymetrix Rat Gene 1.0 ST Array 10925671 - -
          Affymetrix Rat Gene 1.0 ST Array 10925672 - -
          Affymetrix Rat Gene 1.1 ST Array 10925669 - -
          Rat Genome 230 2.0 Array 1382047_PM_at - -
          Rat Genome 230 2.0 Array 1382047_at - -
          Affymetrix HT Rat Focus Array 1382047_PM_at - -
          Affymetrix HT Rat Focus Array 1382047_at - -
          Affymetrix Rat Clariom S Assay PSR0900010394.rn.1 TC0900001190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900010395.rn.1 TC0900001190.rn.1 RTA TC
          Rat Expression Set 230 1382047_PM_at - -
          Rat Expression Set 230 1382047_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925669 - -
          Rat Genome U34 Set rc_AA899888_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21610528 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356262 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0