Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01428192_m1
          See other TET1 GE Assays ›
          Gene Symbol:
          Tet1
          Gene Name:
          tet methylcytosine dioxygenase 1
          Gene Aliases:
          Cxxc6
          Chromosome Location:
          Chr.20: 27359125 - 27438039 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01428192_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tet1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006256398.2 XP_006256460.1 10 - 11 5221 66

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          inner cell mass cell differentiation
          protein O-linked glycosylation
          biological_process
          stem cell population maintenance
          regulation of DNA methylation
          positive regulation of transcription by RNA polymerase II
          DNA demethylation
          negative regulation of DNA methylation-dependent heterochromatin assembly
          regulation of genetic imprinting
          molecular_function
          DNA binding
          iron ion binding
          zinc ion binding
          methylcytosine dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P086073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47162 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086073 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086073 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P086073 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P466577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832849 - -
          Affymetrix Rat Gene 1.0 ST Array 10832856 - -
          Affymetrix Rat Gene 1.0 ST Array 10832853 - -
          Affymetrix Rat Gene 1.0 ST Array 10832851 - -
          Affymetrix Rat Gene 1.0 ST Array 10832855 - -
          Affymetrix Rat Gene 1.0 ST Array 10832852 - -
          Affymetrix Rat Gene 1.0 ST Array 10832858 - -
          Affymetrix Rat Gene 1.0 ST Array 10832857 - -
          Affymetrix Rat Gene 1.0 ST Array 10832848 - -
          Affymetrix Rat Gene 1.1 ST Array 10832847 - -
          Affymetrix Rat Gene 1.0 ST Array 10832847 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20995633 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365132 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0