Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01428915_g1
          See other LOC100360679 GE Assays ›
          Gene Symbol:
          LOC100360679 Rps18
          Gene Name:
          ribosomal protein S18-like
          ribosomal protein S18
          Gene Aliases:
          -
          Ke3
          Chromosome Location:
          Chr.10: 104521700 - 104522235 on Build Rnor_6.0
          Chromosome Location:
          Chr.20: 5441875 - 5445553 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01428915_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100360679
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002727833.4 XP_002727879.1 1 321 73
          Gene Symbol: Rps18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_213557.1 NP_998722.1 3 - 4 329 73
          GenBank mRNA BC126072.1 - 3 - 4 303 8362450 73
          FQ209845.1 - 3 - 4 327 73
          FQ210672.1 - 3 - 4 311 73
          FQ210792.1 - 3 - 4 327 73
          FQ220696.1 - 3 - 4 327 73
          FQ221730.1 - 3 - 4 327 73
          FQ221918.1 - 3 - 4 327 73
          FQ221943.1 - 1 - 2 137 73
          FQ222652.1 - 3 - 4 327 73
          FQ222675.1 - 3 - 4 325 73
          FQ223127.1 - 3 - 4 327 73
          FQ223169.1 - 3 - 4 328 73
          FQ223198.1 - 3 - 4 327 73
          FQ223436.1 - 3 - 4 327 73
          FQ223481.1 - 3 - 4 327 73
          X57529.1 - 3 - 4 321 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100360679
          Click here to find siRNAs for Gene Rps18

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          regulation of translation
          ribosome biogenesis
          structural constituent of ribosome
          rRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P582859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853657 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P582859 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P582859 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P582859 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P582859 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701614262 - -
          Agilent Rat cDNA Microarray (G4105A) 701614262 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828435 - -
          Affymetrix Rat Gene 1.0 ST Array 10749060 - -
          Affymetrix Rat Gene 1.0 ST Array 10870044 - -
          Affymetrix Rat Gene 1.0 ST Array 10870045 - -
          Affymetrix Rat Gene 1.0 ST Array 10828437 - -
          Affymetrix Rat Gene 1.0 ST Array 10828436 - -
          Affymetrix Rat Gene 1.0 ST Array 10828439 - -
          Affymetrix Rat Gene 1.0 ST Array 10828434 - -
          Affymetrix Rat Gene 1.0 ST Array 10828438 - -
          Affymetrix Rat Gene 1.0 ST Array 10749059 - -
          Affymetrix Rat Gene 1.1 ST Array 10870043 - -
          Affymetrix Rat Gene 1.1 ST Array 10749058 - -
          Affymetrix Rat Gene 1.1 ST Array 10828433 - -
          Rat Genome 230 2.0 Array 1388296_PM_at - -
          Rat Genome 230 2.0 Array 1388296_at - -
          Affymetrix Rat Clariom S Assay PSR2000002073.rn.1 TC2000000217.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000002072.rn.1 TC2000000217.rn.1 RTA TC
          Rat Expression Set 230 1388296_PM_at - -
          Rat Expression Set 230 1388296_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870043 - -
          Affymetrix Rat Gene 1.0 ST Array 10749058 - -
          Affymetrix Rat Gene 1.0 ST Array 10828433 - -
          Rat Genome U34 Set X57529cds_s_at - -
          Rat Genome U34 Set rc_AI233220_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22112954 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371064 - -
          RatRef-12 Expression BeadChip ILMN_67184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0