Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01429309_m1
          See other LOC103694870 GE Assays ›
          Gene Symbol:
          LOC103694870 LOC103690052 Slc5a4
          Gene Name:
          low affinity sodium-glucose cotransporter-like
          low affinity sodium-glucose cotransporter-like
          solute carrier family 5 (glucose activated ion channel), member 4
          Gene Aliases:
          -
          -
          Slc5a4a
          Chromosome Location:
          Chr.Un|NW_007906817: 20127 - 51122 on Build Rnor_6.0
          Chromosome Location:
          Chr.20: 13344408 - 13375403 on Build Rnor_6.0
          Chromosome Location:
          Chr.20: 13415039 - 13458110 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01429309_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103694870
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773819.1 XP_008772041.1 9 - 10 1306 79
          Gene Symbol: LOC103690052
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008772964.1 XP_008771186.1 1306 79
          Gene Symbol: Slc5a4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106383.1 NP_001099853.1 14 - 15 1862 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001341.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane transport
          proton transmembrane transport
          transporter activity
          proton transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26830 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167468 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144933 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144933 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144933 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P534490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88018 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90828 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832456 - -
          Affymetrix Rat Gene 1.0 ST Array 10832450 - -
          Affymetrix Rat Gene 1.0 ST Array 10832459 - -
          Affymetrix Rat Gene 1.0 ST Array 10832446 - -
          Affymetrix Rat Gene 1.0 ST Array 10832455 - -
          Affymetrix Rat Gene 1.0 ST Array 10832457 - -
          Affymetrix Rat Gene 1.0 ST Array 10832449 - -
          Affymetrix Rat Gene 1.0 ST Array 10832448 - -
          Affymetrix Rat Gene 1.0 ST Array 10832451 - -
          Affymetrix Rat Gene 1.0 ST Array 10832447 - -
          Affymetrix Rat Gene 1.0 ST Array 10832458 - -
          Affymetrix Rat Gene 1.0 ST Array 10832453 - -
          Affymetrix Rat Gene 1.0 ST Array 10832452 - -
          Affymetrix Rat Gene 1.0 ST Array 10832454 - -
          Affymetrix Rat Gene 1.1 ST Array 10832445 - -
          Affymetrix Rat Clariom S Assay PSR2000011104.rn.1 TC2000001341.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10832445 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21120480 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373559 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0