Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01429339_m1
          See other UBE2G2 GE Assays ›
          Gene Symbol:
          Ube2g2
          Gene Name:
          ubiquitin-conjugating enzyme E2G 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 11700089 - 11721823 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01429339_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2g2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106380.1 NP_001099850.1 5 - 6 457 80
          GenBank mRNA BC166733.1 - 5 - 6 390 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001302.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          protein ubiquitination
          protein N-linked glycosylation via asparagine
          ubiquitin-dependent ERAD pathway
          cellular protein catabolic process
          protein K48-linked ubiquitination
          ubiquitin-protein transferase activity
          ligase activity
          ubiquitin-like protein transferase activity
          ubiquitin protein ligase binding
          identical protein binding
          ubiquitin protein ligase activity
          ubiquitin conjugating enzyme activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082889 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082889 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1023515 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103163 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86815 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832289 - -
          Affymetrix Rat Gene 1.0 ST Array 10832288 - -
          Affymetrix Rat Gene 1.0 ST Array 10832293 - -
          Affymetrix Rat Gene 1.0 ST Array 10832291 - -
          Affymetrix Rat Gene 1.0 ST Array 10832292 - -
          Affymetrix Rat Gene 1.0 ST Array 10832290 - -
          Affymetrix Rat Gene 1.1 ST Array 10832287 - -
          Rat Genome 230 2.0 Array 1380157_at - -
          Rat Genome 230 2.0 Array 1380157_PM_at - -
          Rat Genome 230 2.0 Array 1397274_PM_at - -
          Rat Genome 230 2.0 Array 1371814_at - -
          Rat Genome 230 2.0 Array 1397274_at - -
          Rat Genome 230 2.0 Array 1371814_PM_at - -
          Rat Genome 230 2.0 Array 1388640_PM_at - -
          Rat Genome 230 2.0 Array 1388640_at - -
          Affymetrix HT Rat Focus Array 1371814_PM_at - -
          Affymetrix HT Rat Focus Array 1380157_at - -
          Affymetrix HT Rat Focus Array 1371814_at - -
          Affymetrix HT Rat Focus Array 1380157_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000010712.rn.1 TC2000001302.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000010710.rn.1 TC2000001302.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000010711.rn.1 TC2000001302.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000010713.rn.1 TC2000001302.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000010714.rn.1 TC2000001302.rn.1 RTA TC
          Rat Expression Set 230 1371814_PM_at - -
          Rat Expression Set 230 1371814_at - -
          Rat Expression Set 230 1380157_at - -
          Rat Expression Set 230 1380157_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10832287 - -
          Rat Genome U34 Set rc_AA859722_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21296368 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366286 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline