Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01430032_m1
          See other LOC100910833 GE Assays ›
          Gene Symbol:
          LOC100910833 Setd3
          Gene Name:
          histone-lysine N-methyltransferase setd3-like
          SET domain containing 3
          Gene Aliases:
          -
          RGD1309550
          Chromosome Location:
          Chr.6: 131602143 - 131650422 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 132023128 - 132090070 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01430032_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100910833
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008764908.1 XP_008763130.1 8 - 9 2741 80
          Gene Symbol: Setd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_216781.8 XP_216781.6 11 - 12 1338 80
          GenBank mRNA BC089108.1 - 6 - 7 738 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          histone H3-K36 methylation
          peptidyl-lysine methylation
          positive regulation of transcription by RNA polymerase II
          positive regulation of muscle cell differentiation
          histone H3-K4 methylation
          regulation of nucleic acid-templated transcription
          transcription coactivator activity
          histone methyltransferase activity (H3-K4 specific)
          histone methyltransferase activity (H3-K36 specific)

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138405 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P122988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P122988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P122988 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P122988 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138405 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1018067 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P122988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46708 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892122 - -
          Affymetrix Rat Gene 1.0 ST Array 10892121 - -
          Affymetrix Rat Gene 1.0 ST Array 10892123 - -
          Affymetrix Rat Gene 1.0 ST Array 10892126 - -
          Affymetrix Rat Gene 1.0 ST Array 10892120 - -
          Affymetrix Rat Gene 1.0 ST Array 10892124 - -
          Affymetrix Rat Gene 1.0 ST Array 10892127 - -
          Affymetrix Rat Gene 1.0 ST Array 10892125 - -
          Affymetrix Rat Gene 1.1 ST Array 10892118 - -
          Rat Genome 230 2.0 Array 1372713_PM_at - -
          Rat Genome 230 2.0 Array 1372713_at - -
          Rat Genome 230 2.0 Array 1374913_at - -
          Rat Genome 230 2.0 Array 1374913_PM_at - -
          Affymetrix HT Rat Focus Array 1374913_PM_at - -
          Affymetrix HT Rat Focus Array 1372713_at - -
          Affymetrix HT Rat Focus Array 1372713_PM_at - -
          Affymetrix HT Rat Focus Array 1374913_at - -
          Rat Expression Set 230 1374913_at - -
          Rat Expression Set 230 1374913_PM_at - -
          Rat Expression Set 230 1372713_at - -
          Rat Expression Set 230 1372713_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10892118 - -
          Rat Genome U34 Set rc_AI176484_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20839981 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367758 - -
          RatRef-12 Expression BeadChip ILMN_52975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix