Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01430075_m1
          See other RGD1566401 GE Assays ›
          Gene Symbol:
          RGD1566401
          Gene Name:
          similar to GTL2, imprinted maternally expressed untranslated
          Gene Aliases:
          Gtl2, Meg3
          Chromosome Location:
          Chr.6: 133658913 - 133691130 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01430075_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1566401
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC158675.1 - 3 - 4 793 67

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56776 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886817 - -
          Affymetrix Rat Gene 1.0 ST Array 10886818 - -
          Affymetrix Rat Gene 1.0 ST Array 10886821 - -
          Affymetrix Rat Gene 1.0 ST Array 10886819 - -
          Affymetrix Rat Gene 1.0 ST Array 10886820 - -
          Affymetrix Rat Gene 1.1 ST Array 10886816 - -
          Rat Genome 230 2.0 Array 1377445_PM_at - -
          Rat Genome 230 2.0 Array 1377339_at - -
          Rat Genome 230 2.0 Array 1388357_at - -
          Rat Genome 230 2.0 Array 1377445_at - -
          Rat Genome 230 2.0 Array 1377339_PM_at - -
          Rat Genome 230 2.0 Array 1377008_at - -
          Rat Genome 230 2.0 Array 1377008_PM_at - -
          Rat Genome 230 2.0 Array 1388357_PM_at - -
          Affymetrix HT Rat Focus Array 1377008_at - -
          Affymetrix HT Rat Focus Array 1377339_PM_at - -
          Affymetrix HT Rat Focus Array 1388357_PM_at - -
          Affymetrix HT Rat Focus Array 1377339_at - -
          Affymetrix HT Rat Focus Array 1388357_at - -
          Affymetrix HT Rat Focus Array 1377445_at - -
          Affymetrix HT Rat Focus Array 1377445_PM_at - -
          Affymetrix HT Rat Focus Array 1377008_PM_at - -
          Rat Expression Set 230 1388357_PM_at - -
          Rat Expression Set 230 1388357_at - -
          Rat Expression Set 230 1377008_PM_at - -
          Rat Expression Set 230 1377008_at - -
          Rat Expression Set 230 1377339_at - -
          Rat Expression Set 230 1377339_PM_at - -
          Rat Expression Set 230 1377445_PM_at - -
          Rat Expression Set 230 1377445_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886816 - -
          Rat Genome U34 Set D83349_at - -
          Rat Genome U34 Set rc_AA943010_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356050 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370261 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0