Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01432537_m1
          See other RNASEH2A GE Assays ›
          Gene Symbol:
          Rnaseh2a
          Gene Name:
          ribonuclease H2, subunit A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 26074980 - 26084679 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01432537_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnaseh2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013234.1 NP_001013252.1 8 - 9 1339 63
          XM_008772414.1 XP_008770636.1 8 - 9 869 63
          XM_008772415.1 XP_008770637.1 6 - 7 653 63
          XM_008772416.1 XP_008770638.1 6 - 7 649 63
          GenBank mRNA BC086345.1 - 8 - 9 869 63
          BC086539.1 - 8 - 9 1339 63
          FQ227287.1 - 6 - 7 698 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001215.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnaseh2a

          Gene Ontology Categories:

          Function(s) Process(es)

          mismatch repair
          RNA catabolic process
          RNA phosphodiester bond hydrolysis, endonucleolytic
          RNA binding
          RNA-DNA hybrid ribonuclease activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54468 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10088 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10088 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10088 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10088 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806594 - -
          Affymetrix Rat Gene 1.0 ST Array 10806590 - -
          Affymetrix Rat Gene 1.0 ST Array 10806598 - -
          Affymetrix Rat Gene 1.0 ST Array 10806593 - -
          Affymetrix Rat Gene 1.0 ST Array 10806597 - -
          Affymetrix Rat Gene 1.0 ST Array 10806591 - -
          Affymetrix Rat Gene 1.0 ST Array 10806588 - -
          Affymetrix Rat Gene 1.0 ST Array 10806600 - -
          Affymetrix Rat Gene 1.0 ST Array 10806599 - -
          Affymetrix Rat Gene 1.0 ST Array 10806589 - -
          Affymetrix Rat Gene 1.0 ST Array 10806596 - -
          Affymetrix Rat Gene 1.0 ST Array 10806592 - -
          Affymetrix Rat Gene 1.1 ST Array 10806587 - -
          Rat Genome 230 2.0 Array 1371451_at - -
          Rat Genome 230 2.0 Array 1371451_PM_at - -
          Affymetrix HT Rat Focus Array 1371451_at - -
          Affymetrix HT Rat Focus Array 1371451_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900009927.rn.1 TC1900001215.rn.1 RTA TC
          Rat Expression Set 230 1371451_at - -
          Rat Expression Set 230 1371451_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806587 - -
          Rat Genome U34 Set rc_AA892777_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22255729 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0