Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435155_m1
          See other BLOC1S5 GE Assays ›
          Gene Symbol:
          Bloc1s5
          Gene Name:
          biogenesis of lysosomal organelles complex-1, subunit 5, muted
          Gene Aliases:
          Muted
          Chromosome Location:
          Chr.17: 26808185 - 26833253 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435155_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bloc1s5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107347.1 NP_001100817.1 4 - 5 456 74
          GenBank mRNA BC166529.1 - 4 - 5 382 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000432.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          anterograde axonal transport
          vesicle-mediated transport
          neuron projection development
          melanosome transport
          otolith morphogenesis
          endosome to melanosome transport
          developmental pigmentation
          anterograde synaptic vesicle transport
          positive regulation of pigment cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144607 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17531 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17531 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17531 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17531 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64555 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794688 - -
          Affymetrix Rat Gene 1.0 ST Array 10794687 - -
          Affymetrix Rat Gene 1.0 ST Array 10794689 - -
          Affymetrix Rat Gene 1.0 ST Array 10794690 - -
          Affymetrix Rat Gene 1.0 ST Array 10794686 - -
          Affymetrix Rat Gene 1.1 ST Array 10794685 - -
          Rat Genome 230 2.0 Array 1372227_PM_at - -
          Rat Genome 230 2.0 Array 1372227_at - -
          Affymetrix HT Rat Focus Array 1372227_PM_at - -
          Affymetrix HT Rat Focus Array 1372227_at - -
          Affymetrix Rat Clariom S Assay PSR1700003139.rn.1 TC1700000432.rn.1 RTA TC
          Rat Expression Set 230 1372227_PM_at - -
          Rat Expression Set 230 1372227_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794685 - -
          Rat Genome U34 Set rc_AA818287_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22258076 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530278 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350942 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0