Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435573_g1
          See other ABO GE Assays ›
          Gene Symbol:
          Abo
          Gene Name:
          ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
          Gene Aliases:
          Abo2
          Chromosome Location:
          Chr.3: 4374679 - 4394374 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435573_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160263.1 NP_001153735.1 4 - 5 226 58
          GenBank mRNA AB081652.1 - 4 - 5 270 58
          AY228143.1 - 4 - 5 226 58

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          positive regulation of cell population proliferation
          response to nematode
          response to retinoic acid
          negative regulation of apoptotic process
          cellular oligosaccharide metabolic process
          alpha-1,3-galactosyltransferase activity
          glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase activity
          fucosylgalactoside 3-alpha-galactosyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043027 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118088 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64592 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043027 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043027 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058432 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058432 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043027 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P454351 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843868 - -
          Affymetrix Rat Gene 1.0 ST Array 10834585 - -
          Affymetrix Rat Gene 1.0 ST Array 10843867 - -
          Affymetrix Rat Gene 1.0 ST Array 10834587 - -
          Affymetrix Rat Gene 1.0 ST Array 10843869 - -
          Affymetrix Rat Gene 1.0 ST Array 10834586 - -
          Affymetrix Rat Gene 1.1 ST Array 10834584 - -
          Affymetrix Rat Gene 1.1 ST Array 10843866 - -
          Affymetrix HT Rat Focus Array 1393031_at - -
          Affymetrix HT Rat Focus Array 1393031_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300020754.rn.1 TC0300002739.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300020753.rn.1 TC0300002739.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300020756.rn.1 TC0300002739.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300020755.rn.1 TC0300002739.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10834584 - -
          Affymetrix Rat Gene 1.0 ST Array 10843866 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21257867 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374268 - -
          RatRef-12 Expression BeadChip ILMN_49106 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0