Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01436318_g1
          See other LOC102555453 GE Assays ›
          Gene Symbol:
          LOC102555453 Rpl12
          Gene Name:
          60S ribosomal protein L12-like
          ribosomal protein L12
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.X: 1345540 - 1346247 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 12009578 - 12011952 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01436318_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC102555453
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006256635.2 XP_006256697.1 1 551 117
          Gene Symbol: Rpl12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109198.1 NP_001102668.1 6 - 7 573 117
          GenBank mRNA BC166903.1 - 6 - 7 574 117
          FQ209969.1 - 4 - 5 357 117
          FQ210206.1 - 4 - 5 356 117
          FQ210369.1 - 4 - 5 357 117
          FQ211114.1 - 4 - 5 357 117
          FQ218073.1 - 4 - 5 357 117
          FQ220669.1 - 4 - 5 357 117
          FQ220753.1 - 4 - 5 357 117
          FQ221328.1 - 4 - 5 357 117
          FQ221528.1 - 4 - 5 357 117
          FQ221617.1 - 4 - 5 357 117
          FQ221651.1 - 4 - 5 357 117
          FQ221748.1 - 4 - 5 357 117
          FQ222328.1 - 4 - 5 357 117
          FQ222394.1 - 4 - 5 357 117
          FQ222706.1 - 4 - 5 357 117
          FQ223807.1 - 4 - 5 357 117
          FQ224022.1 - 4 - 5 357 117
          FQ224617.1 - 4 - 5 357 117
          FQ227863.1 - 4 - 5 357 117
          FQ228502.1 - 4 - 5 357 117
          FQ231502.1 - 4 - 5 357 117
          X53504.1 - 6 - 7 566 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P076002 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353220 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076002 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076002 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071825 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076002 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267103 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267106 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700142832 - -
          Agilent Rat cDNA Microarray (G4105A) 700142832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835646 - -
          Affymetrix Rat Gene 1.0 ST Array 10862676 - -
          Affymetrix Rat Gene 1.0 ST Array 10855675 - -
          Affymetrix Rat Gene 1.0 ST Array 10835650 - -
          Affymetrix Rat Gene 1.0 ST Array 10752099 - -
          Affymetrix Rat Gene 1.0 ST Array 10911796 - -
          Affymetrix Rat Gene 1.0 ST Array 10902468 - -
          Affymetrix Rat Gene 1.0 ST Array 10835649 - -
          Affymetrix Rat Gene 1.0 ST Array 10835647 - -
          Affymetrix Rat Gene 1.0 ST Array 10835648 - -
          Affymetrix Rat Gene 1.0 ST Array 10936481 - -
          Affymetrix Rat Gene 1.0 ST Array 10835651 - -
          Affymetrix Rat Gene 1.1 ST Array 10862675 - -
          Affymetrix Rat Gene 1.1 ST Array 10936480 - -
          Affymetrix Rat Gene 1.1 ST Array 10902467 - -
          Affymetrix Rat Gene 1.1 ST Array 10911795 - -
          Affymetrix Rat Gene 1.1 ST Array 10855674 - -
          Affymetrix Rat Gene 1.1 ST Array 10752098 - -
          Affymetrix Rat Gene 1.1 ST Array 10835645 - -
          Rat Genome 230 2.0 Array 1375181_at - -
          Rat Genome 230 2.0 Array 1375181_PM_at - -
          Affymetrix HT Rat Focus Array 1375181_PM_at - -
          Affymetrix HT Rat Focus Array 1375181_at - -
          Affymetrix Rat Clariom S Assay PSR0300003146.rn.1 TC0300000275.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003149.rn.1 TC0300000275.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003151.rn.1 TC0300000275.rn.1 RTA TC
          Rat Expression Set 230 1375181_PM_at - -
          Rat Expression Set 230 1375181_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936480 - -
          Affymetrix Rat Gene 1.0 ST Array 10911795 - -
          Affymetrix Rat Gene 1.0 ST Array 10862675 - -
          Affymetrix Rat Gene 1.0 ST Array 10902467 - -
          Affymetrix Rat Gene 1.0 ST Array 10855674 - -
          Affymetrix Rat Gene 1.0 ST Array 10835645 - -
          Rat Genome U34 Set X53504cds_g_at - -
          Rat Genome U34 Set X53504cds_g_at - -
          Rat Genome U34 Set X53504cds_at - -
          Rat Genome U34 Set X53504cds_at - -
          Rat Genome U34 Set X53504cds_at - -
          Rat Genome U34 Set X53504cds_g_at - -
          Rat Genome U34 Set rc_AI103105_i_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364180 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356720 - -
          RatRef-12 Expression BeadChip ILMN_53063 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0