Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01436721_g1
          See other NDUFAF4 GE Assays ›
          Gene Symbol:
          Ndufaf4
          Gene Name:
          NADH dehydrogenase (ubiquinone) complex I, assembly factor 4
          Gene Aliases:
          Hrpap20
          Chromosome Location:
          Chr.5: 39251370 - 39256542 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01436721_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufaf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198783.1 NP_942078.1 2 - 3 279 93
          GenBank mRNA AF283900.1 - 2 - 3 279 93
          AF283901.1 - 1 - 2 542 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000335.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufaf4

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell population proliferation
          NADH dehydrogenase complex assembly
          mitochondrial respiratory chain complex I assembly
          negative regulation of apoptotic process
          calmodulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P470528 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P122646 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P253940 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P394011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443351 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122646 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P253940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P394011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P470528 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P253940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P470528 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122646 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P394011 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P470528 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P253940 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P122646 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P394011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P394011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P470528 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P253940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46428 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867947 - -
          Affymetrix Rat Gene 1.0 ST Array 10867945 - -
          Affymetrix Rat Gene 1.0 ST Array 10867946 - -
          Affymetrix Rat Gene 1.1 ST Array 10867944 - -
          Rat Genome 230 2.0 Array 1389384_PM_at - -
          Rat Genome 230 2.0 Array 1389384_at - -
          Affymetrix HT Rat Focus Array 1389384_PM_at - -
          Affymetrix HT Rat Focus Array 1389384_at - -
          Affymetrix Rat Clariom S Assay PSR0500002108.rn.1 TC0500000335.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002106.rn.1 TC0500000335.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002107.rn.1 TC0500000335.rn.1 RTA TC
          Rat Expression Set 230 1389384_at - -
          Rat Expression Set 230 1389384_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867944 - -
          Rat Genome U34 Set rc_AA996463_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20865395 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352329 - -
          RatRef-12 Expression BeadChip ILMN_66842 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline