Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01437184_m1
          See other TMEM194B GE Assays ›
          Gene Symbol:
          Tmem194b
          Gene Name:
          transmembrane protein 194B
          Gene Aliases:
          RGD1560421
          Chromosome Location:
          Chr.9: 53713598 - 53732858 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01437184_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem194b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134642.1 NP_001128114.1 7 - 8 961 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001939.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093304 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P600052 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146475 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152091 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P600052 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093304 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152091 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093304 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P600052 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152091 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093304 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P600052 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927831 - -
          Affymetrix Rat Gene 1.0 ST Array 10927837 - -
          Affymetrix Rat Gene 1.0 ST Array 10927829 - -
          Affymetrix Rat Gene 1.0 ST Array 10927833 - -
          Affymetrix Rat Gene 1.0 ST Array 10927836 - -
          Affymetrix Rat Gene 1.0 ST Array 10927832 - -
          Affymetrix Rat Gene 1.0 ST Array 10927834 - -
          Affymetrix Rat Gene 1.0 ST Array 10927830 - -
          Affymetrix Rat Gene 1.0 ST Array 10927835 - -
          Affymetrix Rat Gene 1.1 ST Array 10927828 - -
          Rat Genome 230 2.0 Array 1390553_at - -
          Rat Genome 230 2.0 Array 1390553_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900015941.rn.1 TC0900001939.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900015940.rn.1 TC0900001939.rn.1 RTA TC
          Rat Expression Set 230 1390553_at - -
          Rat Expression Set 230 1390553_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927828 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21302102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0