Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01437728_m1
          See other TGFBRAP1 GE Assays ›
          Gene Symbol:
          Tgfbrap1
          Gene Name:
          transforming growth factor, beta receptor associated protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 49852560 - 49902943 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01437728_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfbrap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106907.1 NP_001100377.1 6 - 7 1670 71
          XM_006244788.2 XP_006244850.1 5 - 6 1268 71
          XM_006244789.2 XP_006244851.1 5 - 6 1221 71
          XM_006244790.2 XP_006244852.1 5 - 6 1227 71
          XM_006244791.2 XP_006244853.1 5 - 6 1247 71
          XM_006244792.2 XP_006244854.1 5 - 6 1165 71
          XM_006244793.2 XP_006244855.1 5 - 6 1990 71
          XM_008767021.1 XP_008765243.1 5 - 6 1306 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001906.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          signal transduction
          regulation of catalytic activity
          transforming growth factor beta receptor binding
          SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390080 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062960 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062960 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062960 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P218241 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67700 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25112 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927686 - -
          Affymetrix Rat Gene 1.0 ST Array 10927685 - -
          Affymetrix Rat Gene 1.0 ST Array 10927681 - -
          Affymetrix Rat Gene 1.0 ST Array 10927682 - -
          Affymetrix Rat Gene 1.0 ST Array 10927687 - -
          Affymetrix Rat Gene 1.0 ST Array 10927683 - -
          Affymetrix Rat Gene 1.0 ST Array 10927690 - -
          Affymetrix Rat Gene 1.0 ST Array 10927691 - -
          Affymetrix Rat Gene 1.0 ST Array 10927684 - -
          Affymetrix Rat Gene 1.0 ST Array 10927689 - -
          Affymetrix Rat Gene 1.0 ST Array 10927688 - -
          Affymetrix Rat Gene 1.1 ST Array 10927680 - -
          Rat Genome 230 2.0 Array 1372231_at - -
          Rat Genome 230 2.0 Array 1372231_PM_at - -
          Affymetrix HT Rat Focus Array 1372231_at - -
          Affymetrix HT Rat Focus Array 1372231_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900015628.rn.1 TC0900001906.rn.1 RTA TC
          Rat Expression Set 230 1372231_at - -
          Rat Expression Set 230 1372231_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927680 - -
          Rat Genome U34 Set rc_AA893532_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21625228 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365395 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0