Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438601_g1
          See other FAM166A GE Assays ›
          Gene Symbol:
          Fam166a
          Gene Name:
          family with sequence similarity 166, member A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 2434367 - 2441706 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438601_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam166a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025694.2 NP_001020865.1 10 - 11 1400 82
          XM_008761596.1 XP_008759818.1 11 - 12 1453 82
          GenBank mRNA BC097396.1 - 10 - 11 1401 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000041.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam166a

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P492245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158201 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492245 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P492245 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492245 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48559 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834151 - -
          Affymetrix Rat Gene 1.0 ST Array 10834152 - -
          Affymetrix Rat Gene 1.0 ST Array 10834153 - -
          Affymetrix Rat Gene 1.0 ST Array 10834154 - -
          Affymetrix Rat Gene 1.0 ST Array 10834147 - -
          Affymetrix Rat Gene 1.0 ST Array 10834155 - -
          Affymetrix Rat Gene 1.0 ST Array 10834149 - -
          Affymetrix Rat Gene 1.0 ST Array 10834145 - -
          Affymetrix Rat Gene 1.0 ST Array 10834146 - -
          Affymetrix Rat Gene 1.0 ST Array 10834148 - -
          Affymetrix Rat Gene 1.0 ST Array 10834150 - -
          Affymetrix Rat Gene 1.0 ST Array 10834156 - -
          Affymetrix Rat Gene 1.1 ST Array 10834144 - -
          Affymetrix Rat Clariom S Assay PSR0300000332.rn.1 TC0300000041.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000333.rn.1 TC0300000041.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10834144 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21715987 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358295 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0