Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438790_g1
          See other GGTA1L1 GE Assays ›
          Gene Symbol:
          Ggta1l1
          Gene Name:
          glycoprotein, alpha-galactosyltransferase 1-like 1
          Gene Aliases:
          Ggta1, Glt6d1
          Chromosome Location:
          Chr.3: 14555916 - 14572863 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438790_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ggta1l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037583.1 NP_001032672.1 4 - 5 289 56
          XM_006234046.2 XP_006234108.1 5 - 6 323 56
          GenBank mRNA AY524048.1 - 4 - 5 289 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002933.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ggta1l1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          metal ion binding
          N-acetyllactosaminide 3-alpha-galactosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1034223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19933 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034223 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034223 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1034223 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59641 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41499 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30268 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844683 - -
          Affymetrix Rat Gene 1.0 ST Array 10844682 - -
          Affymetrix Rat Gene 1.0 ST Array 10844685 - -
          Affymetrix Rat Gene 1.0 ST Array 10844684 - -
          Affymetrix Rat Gene 1.0 ST Array 10844686 - -
          Affymetrix Rat Gene 1.1 ST Array 10844681 - -
          Affymetrix Rat Clariom S Assay PSR0300022890.rn.1 TC0300002933.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10844681 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21565857 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-998b77544-nf2nv:80/100.66.134.75:80.
          git-commit: efd2b0e08ab99238c7874a09efb06372d741ac0a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc