Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441113_m1
          See other ZBTB24 GE Assays ›
          Gene Symbol:
          Zbtb24
          Gene Name:
          zinc finger and BTB domain containing 24
          Gene Aliases:
          Bif1a, ZNF450
          Chromosome Location:
          Chr.20: 46166646 - 46191172 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441113_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001098667.1 NP_001092137.1 5 - 6 1472 96
          XM_008773001.1 XP_008771223.1 5 - 6 3002 96
          XM_008773002.1 XP_008771224.1 5 - 6 1465 96
          XM_008773003.1 XP_008771225.1 5 - 6 3080 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000825.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          DNA-binding transcription factor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P22070 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22070 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22070 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042684 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P22070 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042684 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P22070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830511 - -
          Affymetrix Rat Gene 1.0 ST Array 10830508 - -
          Affymetrix Rat Gene 1.0 ST Array 10830510 - -
          Affymetrix Rat Gene 1.0 ST Array 10830512 - -
          Affymetrix Rat Gene 1.0 ST Array 10830513 - -
          Affymetrix Rat Gene 1.0 ST Array 10830514 - -
          Affymetrix Rat Gene 1.1 ST Array 10830505 - -
          Rat Genome 230 2.0 Array 1385358_at - -
          Rat Genome 230 2.0 Array 1384362_at - -
          Rat Genome 230 2.0 Array 1385358_PM_at - -
          Rat Genome 230 2.0 Array 1384362_PM_at - -
          Affymetrix HT Rat Focus Array 1384362_PM_at - -
          Affymetrix HT Rat Focus Array 1385358_at - -
          Affymetrix HT Rat Focus Array 1384362_at - -
          Affymetrix HT Rat Focus Array 1385358_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000006907.rn.1 TC2000000825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006906.rn.1 TC2000000825.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006908.rn.1 TC2000000825.rn.1 RTA TC
          Rat Expression Set 230 1384362_at - -
          Rat Expression Set 230 1384362_PM_at - -
          Rat Expression Set 230 1385358_PM_at - -
          Rat Expression Set 230 1385358_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830505 - -
          Rat Genome U34 Set rc_AI638999_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21052295 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365313 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0