Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441954_m1
          See other TSPAN17 GE Assays ›
          Gene Symbol:
          Tspan17
          Gene Name:
          tetraspanin 17
          Gene Aliases:
          Fbxo23, GHB-R
          Chromosome Location:
          Chr.17: 10356980 - 10364605 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441954_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tspan17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013138.2 NP_001013156.2 1 - 2 229 77
          XM_006253630.2 XP_006253692.1 1 - 2 262 77
          XM_006253631.2 XP_006253693.1 1 - 2 264 77
          GenBank mRNA AY485933.1 - 1 - 2 223 77
          BC097431.1 - 1 - 2 209 77
          FM038661.1 - 1 - 2 229 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001341.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tspan17

          Gene Ontology Categories:

          Function(s) Process(es)

          protein maturation
          establishment of protein localization to organelle
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P712949 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P538165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P712949 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538165 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538165 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P712949 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P712949 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P538165 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538165 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P712949 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797378 - -
          Affymetrix Rat Gene 1.0 ST Array 10797375 - -
          Affymetrix Rat Gene 1.0 ST Array 10797371 - -
          Affymetrix Rat Gene 1.0 ST Array 10797377 - -
          Affymetrix Rat Gene 1.0 ST Array 10797372 - -
          Affymetrix Rat Gene 1.0 ST Array 10797373 - -
          Affymetrix Rat Gene 1.0 ST Array 10797379 - -
          Affymetrix Rat Gene 1.0 ST Array 10797374 - -
          Affymetrix Rat Gene 1.0 ST Array 10797370 - -
          Affymetrix Rat Gene 1.0 ST Array 10797376 - -
          Affymetrix Rat Gene 1.1 ST Array 10797368 - -
          Rat Genome 230 2.0 Array 1376500_PM_at - -
          Rat Genome 230 2.0 Array 1386614_at - -
          Rat Genome 230 2.0 Array 1386614_PM_at - -
          Rat Genome 230 2.0 Array 1376500_at - -
          Affymetrix HT Rat Focus Array 1386614_PM_at - -
          Affymetrix HT Rat Focus Array 1386614_at - -
          Affymetrix HT Rat Focus Array 1376500_at - -
          Affymetrix HT Rat Focus Array 1376500_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700009353.rn.1 TC1700001341.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009351.rn.1 TC1700001341.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009354.rn.1 TC1700001341.rn.1 RTA TC
          Rat Expression Set 230 1376500_PM_at - -
          Rat Expression Set 230 1386614_PM_at - -
          Rat Expression Set 230 1386614_at - -
          Rat Expression Set 230 1376500_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797368 - -
          Rat Genome U34 Set rc_AI639044_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20943507 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0