Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442395_m1
          See other SMARCD3 GE Assays ›
          Gene Symbol:
          Smarcd3
          Gene Name:
          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 7076711 - 7108334 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442395_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smarcd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011966.1 NP_001011966.1 9 - 10 1038 109
          XM_006235883.2 XP_006235945.1 8 - 9 1135 109
          XM_006235884.2 XP_006235946.1 8 - 9 1135 109
          XM_006235885.2 XP_006235947.1 8 - 9 1099 109
          XM_006235886.2 XP_006235948.1 9 - 10 1122 109
          XM_006235887.2 XP_006235949.1 9 - 10 1086 109
          XM_006235888.2 XP_006235950.1 8 - 9 1135 109
          XM_006235889.2 XP_006235951.1 8 - 9 1099 109
          GenBank mRNA BC085349.1 - 9 - 10 1038 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000097.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smarcd3

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of neuroblast proliferation
          heart morphogenesis
          secondary heart field specification
          cardiac right ventricle formation
          neural retina development
          nucleosome disassembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          positive regulation of G2/M transition of mitotic cell cycle
          muscle cell differentiation
          regulation of protein binding
          positive regulation of transcription, DNA-templated
          positive regulation of smooth muscle cell differentiation
          chromatin binding
          signaling receptor binding
          transcription factor binding
          nuclear receptor binding
          nuclear receptor coactivator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P107851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42503 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107851 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P107851 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034937 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58971 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32448 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852851 - -
          Affymetrix Rat Gene 1.0 ST Array 10852850 - -
          Affymetrix Rat Gene 1.0 ST Array 10852845 - -
          Affymetrix Rat Gene 1.0 ST Array 10852839 - -
          Affymetrix Rat Gene 1.0 ST Array 10852844 - -
          Affymetrix Rat Gene 1.0 ST Array 10852842 - -
          Affymetrix Rat Gene 1.0 ST Array 10852843 - -
          Affymetrix Rat Gene 1.0 ST Array 10852837 - -
          Affymetrix Rat Gene 1.0 ST Array 10852841 - -
          Affymetrix Rat Gene 1.0 ST Array 10852847 - -
          Affymetrix Rat Gene 1.0 ST Array 10852840 - -
          Affymetrix Rat Gene 1.0 ST Array 10852846 - -
          Affymetrix Rat Gene 1.0 ST Array 10852849 - -
          Affymetrix Rat Gene 1.0 ST Array 10852848 - -
          Affymetrix Rat Gene 1.1 ST Array 10852836 - -
          Rat Genome 230 2.0 Array 1371665_PM_at - -
          Rat Genome 230 2.0 Array 1377473_PM_at - -
          Rat Genome 230 2.0 Array 1377473_at - -
          Rat Genome 230 2.0 Array 1371665_at - -
          Affymetrix HT Rat Focus Array 1371665_PM_at - -
          Affymetrix HT Rat Focus Array 1371665_at - -
          Affymetrix Rat Clariom S Assay PSR0400000632.rn.1 TC0400000097.rn.1 RTA TC
          Rat Expression Set 230 1371665_PM_at - -
          Rat Expression Set 230 1371665_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852836 - -
          Rat Genome U34 Set rc_AI145891_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22108890 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376683 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375383 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc