Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442987_m1
          See other PLA2G3 GE Assays ›
          Gene Symbol:
          Pla2g3
          Gene Name:
          phospholipase A2, group III
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 83724933 - 83730614 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442987_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106015.1 NP_001099485.1 6 - 7 1360 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000877.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          acrosome assembly
          sperm axoneme assembly
          lipid catabolic process
          lipoxygenase pathway
          mast cell degranulation
          phosphatidylcholine metabolic process
          cell development
          cilium assembly
          phospholipase A2 activity
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79970 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006948 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006948 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006948 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78436 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773711 - -
          Affymetrix Rat Gene 1.0 ST Array 10773714 - -
          Affymetrix Rat Gene 1.0 ST Array 10773713 - -
          Affymetrix Rat Gene 1.0 ST Array 10773710 - -
          Affymetrix Rat Gene 1.0 ST Array 10773712 - -
          Affymetrix Rat Gene 1.0 ST Array 10773709 - -
          Affymetrix Rat Gene 1.0 ST Array 10773715 - -
          Affymetrix Rat Gene 1.1 ST Array 10773708 - -
          Rat Genome 230 2.0 Array 1384741_at - -
          Rat Genome 230 2.0 Array 1384741_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400006242.rn.1 TC1400000877.rn.1 RTA TC
          Rat Expression Set 230 1384741_PM_at - -
          Rat Expression Set 230 1384741_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773708 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22382224 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372233 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0