Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443665_m1
          See other LOC103693780 GE Assays ›
          Gene Symbol:
          LOC103693780 Ogdh
          Gene Name:
          2-oxoglutarate dehydrogenase, mitochondrial-like
          oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide)
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.14: 86599906 - 86614963 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 86414925 - 86481903 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01443665_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103693780
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008770414.1 XP_008768636.1 1616 80
          Gene Symbol: Ogdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017461.1 NP_001017461.1 20 - 21 2714 80
          XM_006251446.2 XP_006251508.1 20 - 21 2830 80
          XM_006251447.2 XP_006251509.1 20 - 21 2976 80
          XM_006251448.2 XP_006251510.1 20 - 21 2753 80
          GenBank mRNA BC083811.1 - 20 - 21 2714 80
          FQ222842.1 - 20 - 21 2689 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000947.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC103693780
          Click here to find siRNAs for Gene Ogdh

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          glycolytic process
          tricarboxylic acid cycle
          2-oxoglutarate metabolic process
          succinyl-CoA metabolic process
          NADH metabolic process
          cerebellar cortex development
          striatum development
          hippocampus development
          thalamus development
          pyramidal neuron development
          tangential migration from the subventricular zone to the olfactory bulb
          olfactory bulb mitral cell layer development
          oxoglutarate dehydrogenase (succinyl-transferring) activity
          thiamine pyrophosphate binding
          heat shock protein binding
          oxoglutarate dehydrogenase (NAD+) activity
          metal ion binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489392 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117519 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117519 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117519 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P869808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57518 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104228 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93021 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103526 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774050 - -
          Affymetrix Rat Gene 1.0 ST Array 10774054 - -
          Affymetrix Rat Gene 1.0 ST Array 10774064 - -
          Affymetrix Rat Gene 1.0 ST Array 10774051 - -
          Affymetrix Rat Gene 1.0 ST Array 10774046 - -
          Affymetrix Rat Gene 1.0 ST Array 10774048 - -
          Affymetrix Rat Gene 1.0 ST Array 10774067 - -
          Affymetrix Rat Gene 1.0 ST Array 10774069 - -
          Affymetrix Rat Gene 1.0 ST Array 10774052 - -
          Affymetrix Rat Gene 1.0 ST Array 10774053 - -
          Affymetrix Rat Gene 1.0 ST Array 10774047 - -
          Affymetrix Rat Gene 1.0 ST Array 10774056 - -
          Affymetrix Rat Gene 1.0 ST Array 10774066 - -
          Affymetrix Rat Gene 1.0 ST Array 10774061 - -
          Affymetrix Rat Gene 1.0 ST Array 10774059 - -
          Affymetrix Rat Gene 1.0 ST Array 10774062 - -
          Affymetrix Rat Gene 1.0 ST Array 10774065 - -
          Affymetrix Rat Gene 1.0 ST Array 10774060 - -
          Affymetrix Rat Gene 1.0 ST Array 10774068 - -
          Affymetrix Rat Gene 1.0 ST Array 10774057 - -
          Affymetrix Rat Gene 1.0 ST Array 10774058 - -
          Affymetrix Rat Gene 1.0 ST Array 10774063 - -
          Affymetrix Rat Gene 1.0 ST Array 10774055 - -
          Affymetrix Rat Gene 1.1 ST Array 10774045 - -
          Rat Genome 230 2.0 Array 1390020_at - -
          Rat Genome 230 2.0 Array 1390020_PM_at - -
          Rat Genome 230 2.0 Array 1397782_PM_at - -
          Rat Genome 230 2.0 Array 1397782_at - -
          Affymetrix HT Rat Focus Array 1397782_PM_at - -
          Affymetrix HT Rat Focus Array 1397782_at - -
          Affymetrix Rat Clariom S Assay PSR1400006988.rn.1 TC1400000947.rn.1 RTA TC
          Rat Expression Set 230 1390020_PM_at - -
          Rat Expression Set 230 1390020_at - -
          Rat Expression Set 230 1397782_PM_at - -
          Rat Expression Set 230 1397782_at - -
          Affymetrix Rat Gene 1.0 ST Array 10774045 - -
          Rat Genome U34 Set rc_AI105084_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21776850 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354780 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline