Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444786_m1
          See other CD276 GE Assays ›
          Gene Symbol:
          Cd276
          Gene Name:
          Cd276 molecule
          Gene Aliases:
          B7RP-2, B7h3
          Chromosome Location:
          Chr.8: 63319982 - 63350312 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01444786_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd276
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182824.2 NP_877976.1 7 - 8 1063 75
          GenBank mRNA AY190319.1 - 7 - 8 1066 75
          BC100064.1 - 7 - 8 1063 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002658.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd276

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of bone mineralization
          positive regulation of interleukin-2 production
          positive regulation of T cell proliferation
          T cell activation
          negative regulation of T cell proliferation
          positive regulation of osteoblast differentiation
          negative regulation of inflammatory response
          regulation of immune response
          signaling receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P028374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028374 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028374 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P028374 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P473467 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16753 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86236 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102910 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917909 - -
          Affymetrix Rat Gene 1.0 ST Array 10917908 - -
          Affymetrix Rat Gene 1.0 ST Array 10917914 - -
          Affymetrix Rat Gene 1.0 ST Array 10917907 - -
          Affymetrix Rat Gene 1.0 ST Array 10917913 - -
          Affymetrix Rat Gene 1.0 ST Array 10917911 - -
          Affymetrix Rat Gene 1.0 ST Array 10917910 - -
          Affymetrix Rat Gene 1.0 ST Array 10917915 - -
          Affymetrix Rat Gene 1.0 ST Array 10917906 - -
          Affymetrix Rat Gene 1.1 ST Array 10917905 - -
          Rat Genome 230 2.0 Array 1395737_at - -
          Rat Genome 230 2.0 Array 1395737_PM_at - -
          Rat Genome 230 2.0 Array 1374198_PM_at - -
          Rat Genome 230 2.0 Array 1374198_at - -
          Affymetrix HT Rat Focus Array 1395737_PM_at - -
          Affymetrix HT Rat Focus Array 1374198_PM_at - -
          Affymetrix HT Rat Focus Array 1395737_at - -
          Affymetrix HT Rat Focus Array 1374198_at - -
          Affymetrix Rat Clariom S Assay PSR0800021593.rn.1 TC0800002658.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021596.rn.1 TC0800002658.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021595.rn.1 TC0800002658.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021594.rn.1 TC0800002658.rn.1 RTA TC
          Rat Expression Set 230 1374198_PM_at - -
          Rat Expression Set 230 1374198_at - -
          Rat Expression Set 230 1395737_at - -
          Rat Expression Set 230 1395737_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917905 - -
          Rat Genome U34 Set rc_AI009703_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21205854 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360321 - -
          RatRef-12 Expression BeadChip ILMN_61049 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline