Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01445294_g1
          See other CSRNP1 GE Assays ›
          Gene Symbol:
          Csrnp1
          Gene Name:
          cysteine-serine-rich nuclear protein 1
          Gene Aliases:
          Axud1
          Chromosome Location:
          Chr.8: 128659883 - 128673335 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01445294_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csrnp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108786.1 NP_001102256.1 4 - 5 884 113
          XM_006244119.2 XP_006244181.1 4 - 5 1551 113
          XM_006244120.2 XP_006244182.1 4 - 5 1156 113
          XM_006244121.1 XP_006244183.1 4 - 5 3307 113
          XM_006244122.2 XP_006244184.1 4 - 5 901 113
          XM_008766688.1 XP_008764910.1 3 - 4 4224 113
          XM_008766689.1 XP_008764911.1 4 - 5 3168 113

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          post-embryonic development
          positive regulation of transcription by RNA polymerase II
          platelet-derived growth factor receptor signaling pathway
          skeletal system morphogenesis
          roof of mouth development
          face morphogenesis
          DNA-binding transcription activator activity, RNA polymerase II-specific
          DNA-binding transcription factor activity
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P431774 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P284129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704556 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P284129 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431774 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P284129 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431774 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P284129 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P431774 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P284129 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920972 - -
          Affymetrix Rat Gene 1.0 ST Array 10920970 - -
          Affymetrix Rat Gene 1.0 ST Array 10920968 - -
          Affymetrix Rat Gene 1.0 ST Array 10920971 - -
          Affymetrix Rat Gene 1.0 ST Array 10920969 - -
          Affymetrix Rat Gene 1.1 ST Array 10920967 - -
          Rat Genome 230 2.0 Array 1389402_PM_at - -
          Rat Genome 230 2.0 Array 1389402_at - -
          Affymetrix HT Rat Focus Array 1389402_at - -
          Affymetrix HT Rat Focus Array 1389402_PM_at - -
          Rat Expression Set 230 1389402_at - -
          Rat Expression Set 230 1389402_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10920967 - -
          Rat Genome U34 Set rc_AI178746_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22161799 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0