Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01445308_s1
          See other UCN2 GE Assays ›
          Gene Symbol:
          Ucn2
          Gene Name:
          urocortin 2
          Gene Aliases:
          Ucn3
          Chromosome Location:
          Chr.8: 117727216 - 117728765 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01445308_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ucn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133385.2 NP_596876.2 2 787 60
          GenBank mRNA AI454706.1 - 1 60

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ucn2

          Gene Ontology Categories:

          Function(s) Process(es)

          cAMP biosynthetic process
          adenylate cyclase-activating G protein-coupled receptor signaling pathway
          digestion
          cell population proliferation
          negative regulation of gene expression
          cellular response to nutrient levels
          negative regulation of luteinizing hormone secretion
          response to immobilization stress
          tissue regeneration
          negative regulation of follicle-stimulating hormone secretion
          cellular response to glucocorticoid stimulus
          cellular response to hypoxia
          G protein-coupled receptor binding
          signaling receptor binding
          hormone activity
          hormone binding
          corticotropin-releasing hormone receptor 2 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092817 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092817 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092817 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P396842 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10913468 - -
          Affymetrix Rat Gene 1.1 ST Array 10913467 - -
          Rat Genome 230 2.0 Array 1369619_at - -
          Rat Genome 230 2.0 Array 1385450_at - -
          Rat Genome 230 2.0 Array 1385450_PM_at - -
          Rat Genome 230 2.0 Array 1369619_PM_at - -
          Affymetrix HT Rat Focus Array 1369619_at - -
          Affymetrix HT Rat Focus Array 1369619_PM_at - -
          Rat Expression Set 230 1369619_at - -
          Rat Expression Set 230 1369619_PM_at - -
          Rat Expression Set 230 1385450_at - -
          Rat Expression Set 230 1385450_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10913467 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22132579 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369080 - -
          RatRef-12 Expression BeadChip ILMN_69751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0