Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01445737_m1
          See other MRAP2 GE Assays ›
          Gene Symbol:
          Mrap2
          Gene Name:
          melanocortin 2 receptor accessory protein 2
          Gene Aliases:
          RGD1309873
          Chromosome Location:
          Chr.8: 94801336 - 94858192 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01445737_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108774.1 NP_001102244.1 4 - 5 413 122
          XM_006243487.2 XP_006243549.1 5 - 6 693 122
          XM_006243488.2 XP_006243550.1 4 - 5 548 122
          XM_008766452.1 XP_008764674.1 5 - 6 681 122
          XM_008766453.1 XP_008764675.1 4 - 5 581 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001247.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          energy reserve metabolic process
          feeding behavior
          protein localization to cell surface
          energy homeostasis
          corticotropin hormone receptor binding
          type 3 melanocortin receptor binding
          type 4 melanocortin receptor binding
          type 5 melanocortin receptor binding
          identical protein binding
          type 1 melanocortin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20380 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022106 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022106 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022106 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P112546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94915 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10912083 - -
          Affymetrix Rat Gene 1.0 ST Array 10912084 - -
          Affymetrix Rat Gene 1.0 ST Array 10912085 - -
          Affymetrix Rat Gene 1.0 ST Array 10912081 - -
          Affymetrix Rat Gene 1.0 ST Array 10912082 - -
          Affymetrix Rat Gene 1.1 ST Array 10912080 - -
          Rat Genome 230 2.0 Array 1378714_at - -
          Rat Genome 230 2.0 Array 1378714_PM_at - -
          Affymetrix HT Rat Focus Array 1378714_at - -
          Affymetrix HT Rat Focus Array 1378714_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800009151.rn.1 TC0800001247.rn.1 RTA TC
          Rat Expression Set 230 1378714_at - -
          Rat Expression Set 230 1378714_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10912080 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21544393 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354720 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0