Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447237_g1
          See other GMPPB GE Assays ›
          Gene Symbol:
          Gmppb
          Gene Name:
          GDP-mannose pyrophosphorylase B
          Gene Aliases:
          RGD1560458
          Chromosome Location:
          Chr.8: 116826234 - 116856159 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447237_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gmppb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108781.1 NP_001102251.1 8 - 9 1185 55
          XM_006243811.2 XP_006243873.1 9 - 10 1482 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003659.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biosynthetic process
          nucleotidyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1045834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624452 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155315 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045834 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045834 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1045834 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36388 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10913148 - -
          Affymetrix Rat Gene 1.0 ST Array 10913147 - -
          Affymetrix Rat Gene 1.0 ST Array 10913152 - -
          Affymetrix Rat Gene 1.0 ST Array 10913150 - -
          Affymetrix Rat Gene 1.0 ST Array 10913146 - -
          Affymetrix Rat Gene 1.0 ST Array 10913143 - -
          Affymetrix Rat Gene 1.0 ST Array 10913149 - -
          Affymetrix Rat Gene 1.0 ST Array 10913145 - -
          Affymetrix Rat Gene 1.0 ST Array 10913144 - -
          Affymetrix Rat Gene 1.1 ST Array 10913141 - -
          Rat Genome 230 2.0 Array 1392701_PM_at - -
          Rat Genome 230 2.0 Array 1392701_at - -
          Rat Genome 230 2.0 Array 1395329_at - -
          Rat Genome 230 2.0 Array 1395329_PM_at - -
          Affymetrix HT Rat Focus Array 1392701_PM_at - -
          Affymetrix HT Rat Focus Array 1392701_at - -
          Affymetrix HT Rat Focus Array 1395329_at - -
          Affymetrix HT Rat Focus Array 1395329_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800029973.rn.1 TC0800003659.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800029969.rn.1 TC0800003659.rn.1 RTA TC
          Rat Expression Set 230 1392701_at - -
          Rat Expression Set 230 1392701_PM_at - -
          Rat Expression Set 230 1395329_PM_at - -
          Rat Expression Set 230 1395329_at - -
          Affymetrix Rat Gene 1.0 ST Array 10913141 - -
          Rat Genome U34 Set rc_AA956285_at - -
          Rat Genome U34 Set rc_AA859302_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350546 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0