Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447987_m1
          See other MTNR1B GE Assays ›
          Gene Symbol:
          Mtnr1b
          Gene Name:
          melatonin receptor 1B
          Gene Aliases:
          Mel1b-r
          Chromosome Location:
          Chr.8: 14359270 - 14373894 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447987_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtnr1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100641.1 NP_001094111.1 1 - 2 340 63
          GenBank mRNA AB377275.1 - 1 - 2 340 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001974.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          glucose homeostasis
          positive regulation of circadian rhythm
          camera-type eye development
          negative regulation of neuron apoptotic process
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of insulin secretion
          regulation of insulin secretion
          negative regulation of cytosolic calcium ion concentration
          negative regulation of transmission of nerve impulse
          positive regulation of transmission of nerve impulse
          regulation of neuronal action potential
          negative regulation of delayed rectifier potassium channel activity
          molecular_function
          melatonin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21448 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157078 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157078 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157078 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P154008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P240912 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915125 - -
          Affymetrix Rat Gene 1.0 ST Array 10915126 - -
          Affymetrix Rat Gene 1.1 ST Array 10915124 - -
          Affymetrix Rat Clariom S Assay PSR0800015848.rn.1 TC0800001974.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800015849.rn.1 TC0800001974.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800015847.rn.1 TC0800001974.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10915124 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21738720 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373099 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0