Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01448875_m1
          See other NEWGENE_1306455 GE Assays ›
          Gene Symbol:
          NEWGENE_1306455 Impad1
          Gene Name:
          inositol monophosphatase domain containing 1
          inositol monophosphatase domain containing 1
          Gene Aliases:
          Impad1
          IMP 3, RGD1306455, gPAPP
          Chromosome Location:
          Chr.5: 17636848 - 17663771 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 90191119 - 90218013 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01448875_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NEWGENE_1306455
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763564.1 XP_008761786.1 3 - 4 900 75
          Gene Symbol: Impad1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008772.2 NP_001008772.2 3 - 4 856 75
          GenBank mRNA AY321340.1 - 1053 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          endochondral ossification
          chondrocyte development
          inositol biosynthetic process
          biological_process
          post-embryonic development
          dephosphorylation
          chondroitin sulfate metabolic process
          embryonic digit morphogenesis
          phosphatidylinositol phosphate biosynthetic process
          molecular_function
          3'-nucleotidase activity
          3'(2'),5'-bisphosphate nucleotidase activity
          inositol monophosphate 1-phosphatase activity
          metal ion binding
          inositol monophosphate 3-phosphatase activity
          inositol monophosphate 4-phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104454 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1034522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226328 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034522 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034522 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1034522 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104454 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139851 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034522 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.1 ST Array 10875311 - -
          Rat Genome 230 2.0 Array 1373367_PM_at - -
          Rat Genome 230 2.0 Array 1381144_PM_at - -
          Rat Genome 230 2.0 Array 1395036_PM_at - -
          Rat Genome 230 2.0 Array 1381144_at - -
          Rat Genome 230 2.0 Array 1373367_at - -
          Rat Genome 230 2.0 Array 1398652_PM_at - -
          Rat Genome 230 2.0 Array 1395036_at - -
          Rat Genome 230 2.0 Array 1398652_at - -
          Affymetrix HT Rat Focus Array 1395036_at - -
          Affymetrix HT Rat Focus Array 1373367_PM_at - -
          Affymetrix HT Rat Focus Array 1398652_at - -
          Affymetrix HT Rat Focus Array 1398652_PM_at - -
          Affymetrix HT Rat Focus Array 1373367_at - -
          Affymetrix HT Rat Focus Array 1395036_PM_at - -
          Rat Expression Set 230 1398652_PM_at - -
          Rat Expression Set 230 1395036_at - -
          Rat Expression Set 230 1395036_PM_at - -
          Rat Expression Set 230 1398652_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20883355 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355156 - -
          RatRef-12 Expression BeadChip ILMN_54093 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0