Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449054_g1
          See other QDPR GE Assays ›
          Gene Symbol:
          Qdpr
          Gene Name:
          quinoid dihydropteridine reductase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 70164682 - 70178284 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449054_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Qdpr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022390.1 NP_071785.1 6 - 7 670 66
          GenBank mRNA BC072536.1 - 6 - 7 683 66
          FQ209462.1 - 4 - 5 413 66
          FQ213843.1 - 6 - 7 699 66
          J03481.1 - 6 - 7 670 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Qdpr

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          L-phenylalanine catabolic process
          tetrahydrobiopterin biosynthetic process
          response to aluminum ion
          response to lead ion
          response to glucagon
          cellular response to xenobiotic stimulus
          6,7-dihydropteridine reductase activity
          protein homodimerization activity
          NADPH binding
          NADH binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494073 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93777 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12219 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12219 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12219 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12219 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700514177 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75554 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36161 - -
          Agilent Rat cDNA Microarray (G4105A) 700514177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773084 - -
          Affymetrix Rat Gene 1.0 ST Array 10773081 - -
          Affymetrix Rat Gene 1.0 ST Array 10773082 - -
          Affymetrix Rat Gene 1.0 ST Array 10773086 - -
          Affymetrix Rat Gene 1.0 ST Array 10773087 - -
          Affymetrix Rat Gene 1.0 ST Array 10773083 - -
          Affymetrix Rat Gene 1.0 ST Array 10773085 - -
          Affymetrix Rat Gene 1.1 ST Array 10773080 - -
          Rat Genome 230 2.0 Array 1367695_PM_at - -
          Rat Genome 230 2.0 Array 1367695_at - -
          Affymetrix HT Rat Focus Array 1367695_at - -
          Affymetrix HT Rat Focus Array 1367695_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400004834.rn.1 TC1400000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004833.rn.1 TC1400000706.rn.1 RTA TC
          Rat Expression Set 230 1367695_PM_at - -
          Rat Expression Set 230 1367695_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773080 - -
          Rat Genome U34 Set J03481mRNA_at - -
          Rat Genome U34 Set J03481mRNA_g_at - -
          Rat Genome U34 Set J03481mRNA_at - -
          Rat Genome U34 Set J03481mRNA_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21675165 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348866 - -
          RatRef-12 Expression BeadChip ILMN_62820 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline