Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449593_m1
          See other LHX2 GE Assays ›
          Gene Symbol:
          Lhx2
          Gene Name:
          LIM homeobox 2
          Gene Aliases:
          LH2A, Lh-2
          Chromosome Location:
          Chr.3: 22640435 - 22658447 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449593_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lhx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106571.1 NP_001100041.1 1 - 2 184 66
          XM_006234072.2 XP_006234134.1 1 - 2 294 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000455.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          hair follicle development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          nervous system development
          axon guidance
          brain development
          mesoderm development
          dorsal/ventral pattern formation
          telencephalon development
          olfactory bulb development
          telencephalon regionalization
          cerebral cortex development
          neurogenesis
          neuron differentiation
          maintenance of epithelial cell apical/basal polarity
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          anatomical structure formation involved in morphogenesis
          axon extension
          retina development in camera-type eye
          negative regulation of transcription regulatory region DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          chromatin binding
          zinc ion binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P240867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30034 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P240867 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P240867 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P240867 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P240867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73978 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835961 - -
          Affymetrix Rat Gene 1.0 ST Array 10835960 - -
          Affymetrix Rat Gene 1.0 ST Array 10835963 - -
          Affymetrix Rat Gene 1.0 ST Array 10835962 - -
          Affymetrix Rat Gene 1.0 ST Array 10835959 - -
          Affymetrix Rat Gene 1.1 ST Array 10835958 - -
          Affymetrix Rat Clariom S Assay PSR0300003932.rn.1 TC0300000455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003934.rn.1 TC0300000455.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10835958 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21420059 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0