Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450729_g1
          See other NDUFS8 GE Assays ›
          Gene Symbol:
          Ndufs8
          Gene Name:
          NADH dehydrogenase (ubiquinone) Fe-S protein 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 219141289 - 219144610 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450729_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufs8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106322.2 NP_001099792.1 6 - 7 547 95
          XM_006230731.1 XP_006230793.1 6 - 7 758 95
          GenBank mRNA BC158791.1 - 6 - 7 547 95
          FQ213239.1 - 6 - 7 758 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007043.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to oxidative stress
          biological_process
          mitochondrial respiratory chain complex I assembly
          molecular_function
          NADH dehydrogenase activity
          NADH dehydrogenase (ubiquinone) activity
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P132522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P132522 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P132522 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P132522 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P132523 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P132522 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19433 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727403 - -
          Affymetrix Rat Gene 1.0 ST Array 10727401 - -
          Affymetrix Rat Gene 1.0 ST Array 10727402 - -
          Affymetrix Rat Gene 1.0 ST Array 10727399 - -
          Affymetrix Rat Gene 1.0 ST Array 10727400 - -
          Affymetrix Rat Gene 1.0 ST Array 10727404 - -
          Affymetrix Rat Gene 1.1 ST Array 10727398 - -
          Rat Genome 230 2.0 Array 1388326_PM_at - -
          Rat Genome 230 2.0 Array 1388326_at - -
          Affymetrix HT Rat Focus Array 1388326_at - -
          Affymetrix HT Rat Focus Array 1388326_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100055936.rn.1 TC0100007043.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055935.rn.1 TC0100007043.rn.1 RTA TC
          Rat Expression Set 230 1388326_at - -
          Rat Expression Set 230 1388326_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727398 - -
          Rat Genome U34 Set rc_AA799479_at - -
          Rat Genome U34 Set rc_AA851379_at - -
          Rat Genome U34 Set AA685112_at - -
          Rat Genome U34 Set rc_AA799479_g_at - -
          Rat Genome U34 Set rc_AA799479_g_at - -
          Rat Genome U34 Set rc_AA851379_at - -
          Rat Genome U34 Set rc_AA799479_at - -
          Rat Genome U34 Set AA685112_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21432557 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0