Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451297_g1
          See other MINPP1 GE Assays ›
          Gene Symbol:
          Minpp1
          Gene Name:
          multiple inositol-polyphosphate phosphatase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 251045374 - 251071045 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451297_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Minpp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019263.1 NP_062136.1 4 - 5 1053 61
          XM_008760326.1 XP_008758548.1 4 - 5 630 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003484.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          dephosphorylation
          phosphatidylinositol-mediated signaling
          acid phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
          inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
          bisphosphoglycerate 3-phosphatase activity
          inositol hexakisphosphate 2-phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082559 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082559 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P349050 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700516749 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701093848 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70367 - -
          Agilent Rat cDNA Microarray (G4105A) 700516749 - -
          Agilent Rat cDNA Microarray (G4105A) 701093848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719149 - -
          Affymetrix Rat Gene 1.0 ST Array 10714817 - -
          Affymetrix Rat Gene 1.1 ST Array 10714814 - -
          Affymetrix Rat Gene 1.1 ST Array 10719148 - -
          Rat Genome 230 2.0 Array 1388126_PM_at - -
          Rat Genome 230 2.0 Array 1388126_at - -
          Affymetrix HT Rat Focus Array 1388126_PM_at - -
          Affymetrix HT Rat Focus Array 1388126_at - -
          Affymetrix Rat Clariom S Assay PSR0100028285.rn.1 TC0100003484.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028284.rn.1 TC0100003484.rn.1 RTA TC
          Rat Expression Set 230 1388126_at - -
          Rat Expression Set 230 1388126_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719148 - -
          Affymetrix Rat Gene 1.0 ST Array 10714814 - -
          Rat Genome U34 Set AF012714_at - -
          Rat Genome U34 Set rc_AI111401_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21296112 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358344 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356070 - -
          RatRef-12 Expression BeadChip ILMN_51516 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0