Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451318_m1
          See other RBP4 GE Assays ›
          Gene Symbol:
          Rbp4
          Gene Name:
          retinol binding protein 4, plasma
          Gene Aliases:
          RBPA
          Chromosome Location:
          Chr.1: 256806476 - 256813678 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451318_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013162.1 NP_037294.1 4 - 5 402 94
          GenBank mRNA BC167099.1 - 4 - 5 403 94
          CO561550.1 - 4 - 5 402 94
          DV725071.1 - 2 - 3 201 94
          M10934.1 - 3 - 4 265 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007651.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          positive regulation of immunoglobulin production
          gluconeogenesis
          transport
          spermatogenesis
          heart development
          male gonad development
          maintenance of gastrointestinal epithelium
          lung development
          positive regulation of insulin secretion
          response to retinoic acid
          response to insulin
          retinol transport
          retinol metabolic process
          retinal metabolic process
          glucose homeostasis
          response to ethanol
          embryonic organ morphogenesis
          embryonic skeletal system development
          cardiac muscle tissue development
          female genitalia morphogenesis
          detection of light stimulus involved in visual perception
          vitamin transport
          retina development in camera-type eye
          negative regulation of cardiac muscle cell proliferation
          embryonic retina morphogenesis in camera-type eye
          uterus development
          vagina development
          urinary bladder development
          heart trabecula formation
          transporter activity
          retinal binding
          retinol binding
          retinol transmembrane transporter activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121085 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058087 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P264611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729976 - -
          Affymetrix Rat Gene 1.0 ST Array 10729971 - -
          Affymetrix Rat Gene 1.0 ST Array 10729975 - -
          Affymetrix Rat Gene 1.0 ST Array 10729974 - -
          Affymetrix Rat Gene 1.0 ST Array 10729973 - -
          Affymetrix Rat Gene 1.0 ST Array 10729972 - -
          Affymetrix Rat Gene 1.1 ST Array 10729970 - -
          Rat Genome 230 2.0 Array 1371762_at - -
          Rat Genome 230 2.0 Array 1371762_PM_at - -
          Affymetrix HT Rat Focus Array 1371762_at - -
          Affymetrix HT Rat Focus Array 1371762_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100061392.rn.1 TC0100007651.rn.1 RTA TC
          Rat Expression Set 230 1371762_PM_at - -
          Rat Expression Set 230 1371762_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729970 - -
          Rat Genome U34 Set K03045cds_r_at - -
          Rat Genome U34 Set M10934_s_at - -
          Rat Genome U34 Set K03045cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21329126 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354144 - -

          Back To Top
           

          Citations (1)

          Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
          Authors
          Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
          Journal
          J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 UTC 2009)
          Volume (Year)
          201 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0