Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453157_m1
          See other GBA3 GE Assays ›
          Gene Symbol:
          Gba3
          Gene Name:
          glucosidase, beta, acid 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 64390793 - 64535212 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453157_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gba3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106010.1 NP_001099480.1 2 - 3 896 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001921.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glycoside catabolic process
          glycosylceramide catabolic process
          hydrolase activity, hydrolyzing O-glycosyl compounds
          beta-galactosidase activity
          beta-glucosidase activity
          glycosylceramidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66006 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75176 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143400 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143400 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143400 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P746647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25844 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39947 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71023 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777117 - -
          Affymetrix Rat Gene 1.0 ST Array 10777118 - -
          Affymetrix Rat Gene 1.0 ST Array 10777119 - -
          Affymetrix Rat Gene 1.0 ST Array 10777120 - -
          Affymetrix Rat Gene 1.1 ST Array 10777116 - -
          Rat Genome 230 2.0 Array 1398628_PM_at - -
          Rat Genome 230 2.0 Array 1398628_at - -
          Affymetrix HT Rat Focus Array 1398628_at - -
          Affymetrix HT Rat Focus Array 1398628_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400013785.rn.1 TC1400001921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013780.rn.1 TC1400001921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013781.rn.1 TC1400001921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013784.rn.1 TC1400001921.rn.1 RTA TC
          Rat Expression Set 230 1398628_at - -
          Rat Expression Set 230 1398628_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777116 - -
          Rat Genome U34 Set rc_AI059960_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21729324 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366951 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0