Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453449_g1
          See other SELK GE Assays ›
          Gene Symbol:
          Selk
          Gene Name:
          selenoprotein K
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 6035926 - 6044240 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453449_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Selk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207589.3 NP_997472.2 3 - 4 283 117
          GenBank mRNA AY319924.1 - 3 - 4 206 117
          BC088086.1 - 3 - 4 227 117
          CF979418.1 - 3 - 4 287 117
          FQ217552.1 - 1 - 2 141 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002111.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Selk

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          response to oxidative stress
          macrophage derived foam cell differentiation
          protein palmitoylation
          positive regulation of cell migration
          positive regulation of T cell proliferation
          respiratory burst after phagocytosis
          regulation of protein transport
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          positive regulation of T cell migration
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1030897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420408 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1030897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1030897 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1030897 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P197783 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1030897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786420 - -
          Affymetrix Rat Gene 1.0 ST Array 10786418 - -
          Affymetrix Rat Gene 1.0 ST Array 10786419 - -
          Affymetrix Rat Gene 1.0 ST Array 10918500 - -
          Affymetrix Rat Gene 1.0 ST Array 10786421 - -
          Affymetrix Rat Gene 1.1 ST Array 10786417 - -
          Affymetrix Rat Gene 1.1 ST Array 10918499 - -
          Rat Genome 230 2.0 Array 1371783_at - -
          Rat Genome 230 2.0 Array 1371783_PM_at - -
          Affymetrix HT Rat Focus Array 1371783_PM_at - -
          Affymetrix HT Rat Focus Array 1371783_at - -
          Affymetrix Rat Clariom S Assay PSR1600015727.rn.1 TC1600002111.rn.1 RTA TC
          Rat Expression Set 230 1371783_PM_at - -
          Rat Expression Set 230 1371783_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918499 - -
          Affymetrix Rat Gene 1.0 ST Array 10786417 - -
          Rat Genome U34 Set rc_AI009936_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350707 - -
          RatRef-12 Expression BeadChip ILMN_58092 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0