Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454176_m1
          See other MS4A6BL GE Assays ›
          Gene Symbol:
          Ms4a6bl LOC100912573
          Gene Name:
          membrane-spanning 4-domains, subfamily A, member 6B-like
          membrane-spanning 4-domains subfamily A member 6B-like
          Gene Aliases:
          Ms4a6b
          -
          Chromosome Location:
          Chr.1: 227240419 - 227252244 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 227757445 - 227769336 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01454176_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ms4a6bl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006975.1 NP_001006976.1 5 - 6 659 73
          GenBank mRNA BC083691.1 - 5 - 6 659 73
          FQ230476.1 - 663 73
          Gene Symbol: LOC100912573
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006231109.2 XP_006231171.1 711 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003216.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ms4a6bl
          Click here to find siRNAs for Gene LOC100912573

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17697 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17697 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17697 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33344 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728928 - -
          Affymetrix Rat Gene 1.0 ST Array 10728929 - -
          Affymetrix Rat Gene 1.0 ST Array 10728926 - -
          Affymetrix Rat Gene 1.0 ST Array 10728925 - -
          Affymetrix Rat Gene 1.0 ST Array 10728927 - -
          Affymetrix Rat Gene 1.1 ST Array 10728924 - -
          Rat Genome 230 2.0 Array 1390510_PM_at - -
          Rat Genome 230 2.0 Array 1390510_at - -
          Affymetrix HT Rat Focus Array 1390510_at - -
          Affymetrix HT Rat Focus Array 1390510_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100026627.rn.1 TC0100003216.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026628.rn.1 TC0100003216.rn.1 RTA TC
          Rat Expression Set 230 1390510_PM_at - -
          Rat Expression Set 230 1390510_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728924 - -
          Rat Genome U34 Set rc_AI012739_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21716320 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_67686 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline