Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454357_m1
          See other LSP1 GE Assays ›
          Gene Symbol:
          Lsp1
          Gene Name:
          lymphocyte-specific protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 215628748 - 215662505 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454357_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025420.1 NP_001020591.1 2 - 3 259 80
          XM_006230815.2 XP_006230877.1 3 - 4 921 80
          XM_006230817.2 XP_006230879.1 2 - 3 296 80
          XM_006230818.1 XP_006230880.1 2 - 3 354 80
          XM_008760146.1 XP_008758368.1 2 - 3 334 80
          XM_008760147.1 XP_008758369.1 2 - 3 583 80
          GenBank mRNA BC097966.1 - 2 - 3 233 80
          CO556739.1 - 2 - 3 259 80
          FQ230230.1 - 2 - 3 240 80
          FQ233963.1 - 2 - 3 249 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002921.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          chemotaxis
          defense response
          signal transduction

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P013226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74554 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013226 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013226 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P013226 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P621642 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80971 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80929 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712480 - -
          Affymetrix Rat Gene 1.0 ST Array 10712487 - -
          Affymetrix Rat Gene 1.0 ST Array 10712485 - -
          Affymetrix Rat Gene 1.0 ST Array 10712486 - -
          Affymetrix Rat Gene 1.0 ST Array 10712482 - -
          Affymetrix Rat Gene 1.0 ST Array 10712481 - -
          Affymetrix Rat Gene 1.0 ST Array 10712489 - -
          Affymetrix Rat Gene 1.0 ST Array 10712483 - -
          Affymetrix Rat Gene 1.0 ST Array 10712488 - -
          Affymetrix Rat Gene 1.0 ST Array 10712478 - -
          Affymetrix Rat Gene 1.0 ST Array 10712484 - -
          Affymetrix Rat Gene 1.1 ST Array 10712477 - -
          Rat Genome 230 2.0 Array 1388673_PM_at - -
          Rat Genome 230 2.0 Array 1388673_at - -
          Affymetrix HT Rat Focus Array 1388673_PM_at - -
          Affymetrix HT Rat Focus Array 1388673_at - -
          Affymetrix Rat Clariom S Assay PSR0100023437.rn.1 TC0100002921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023452.rn.1 TC0100002921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023451.rn.1 TC0100002921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023453.rn.1 TC0100002921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023454.rn.1 TC0100002921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023456.rn.1 TC0100002921.rn.1 RTA TC
          Rat Expression Set 230 1388673_PM_at - -
          Rat Expression Set 230 1388673_at - -
          Affymetrix Rat Gene 1.0 ST Array 10712477 - -
          Rat Genome U34 Set rc_AI012109_at - -
          Rat Genome U34 Set rc_AI103247_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20990022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline