Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454505_m1
          See other NDUFV1 GE Assays ›
          Gene Symbol:
          Ndufv1
          Gene Name:
          NADH dehydrogenase (ubiquinone) flavoprotein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 219254293 - 219259328 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454505_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufv1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006972.1 NP_001006973.1 1 - 2 85 61
          GenBank mRNA BC083709.1 - 1 - 2 85 61
          FQ214688.1 - 1 - 2 92 61
          FQ217076.1 - 1 - 2 143 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007048.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufv1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          NADH dehydrogenase (ubiquinone) activity
          FMN binding
          NAD binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140691 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140691 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140691 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16415 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62841 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727423 - -
          Affymetrix Rat Gene 1.0 ST Array 10727420 - -
          Affymetrix Rat Gene 1.0 ST Array 10727424 - -
          Affymetrix Rat Gene 1.0 ST Array 10727428 - -
          Affymetrix Rat Gene 1.0 ST Array 10727426 - -
          Affymetrix Rat Gene 1.0 ST Array 10727427 - -
          Affymetrix Rat Gene 1.0 ST Array 10727422 - -
          Affymetrix Rat Gene 1.0 ST Array 10727425 - -
          Affymetrix Rat Gene 1.0 ST Array 10727419 - -
          Affymetrix Rat Gene 1.0 ST Array 10727421 - -
          Affymetrix Rat Gene 1.1 ST Array 10727418 - -
          Rat Genome 230 2.0 Array 1371416_at - -
          Rat Genome 230 2.0 Array 1371416_PM_at - -
          Affymetrix HT Rat Focus Array 1371416_at - -
          Affymetrix HT Rat Focus Array 1371416_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100055991.rn.1 TC0100007048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055994.rn.1 TC0100007048.rn.1 RTA TC
          Rat Expression Set 230 1371416_PM_at - -
          Rat Expression Set 230 1371416_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727418 - -
          Rat Genome U34 Set rc_AA901223_s_at - -
          Rat Genome U34 Set rc_AA901223_i_at - -
          Rat Genome U34 Set rc_AI103960_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21978473 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365082 - -
          RatRef-12 Expression BeadChip ILMN_49421 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0