Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031527.1 | NP_113715.1 | 6 - 7 | 951 | 82 | |
GenBank mRNA | BC070517.1 | - | 6 - 7 | 931 | 82 | |
D00859.1 | - | 6 - 7 | 951 | 82 | ||
D90163.1 | - | 6 - 7 | 935 | 82 | ||
FQ221064.1 | - | 1 - 2 | 82 | |||
FQ224860.1 | - | 6 - 7 | 941 | 82 | ||
FQ231613.1 | - | 6 - 7 | 960 | 82 | ||
FQ232496.1 | - | 6 - 7 | 959 | 82 | ||
FQ233040.1 | - | 6 - 7 | 951 | 82 | ||
FQ235125.1 | - | 6 - 7 | 953 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |