Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454709_m1
          See other AKT1S1 GE Assays ›
          Gene Symbol:
          Akt1s1
          Gene Name:
          AKT1 substrate 1 (proline-rich)
          Gene Aliases:
          PRAS40
          Chromosome Location:
          Chr.1: 100845445 - 100851961 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454709_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akt1s1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106259.3 NP_001099729.2 4 - 5 748 79
          XM_006229003.2 XP_006229065.1 4 - 5 1224 79
          XM_006229004.2 XP_006229066.1 4 - 5 924 79
          XM_006229005.2 XP_006229067.1 4 - 5 900 79
          XM_008759335.1 XP_008757557.1 3 - 4 477 79
          GenBank mRNA EV773331.1 - 3 - 4 664 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001467.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          biological_process
          negative regulation of TOR signaling
          regulation of neuron apoptotic process
          negative regulation of cell size
          neurotrophin TRK receptor signaling pathway
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P180255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P180255 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P180255 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P180255 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P180255 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P666786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95094 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54634 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54389 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706649 - -
          Affymetrix Rat Gene 1.0 ST Array 10706646 - -
          Affymetrix Rat Gene 1.0 ST Array 10706647 - -
          Affymetrix Rat Gene 1.0 ST Array 10706648 - -
          Affymetrix Rat Gene 1.1 ST Array 10706644 - -
          Rat Genome 230 2.0 Array 1375766_PM_at - -
          Rat Genome 230 2.0 Array 1375766_at - -
          Rat Genome 230 2.0 Array 1372879_PM_at - -
          Rat Genome 230 2.0 Array 1375117_at - -
          Rat Genome 230 2.0 Array 1372879_at - -
          Rat Genome 230 2.0 Array 1375117_PM_at - -
          Affymetrix HT Rat Focus Array 1372879_at - -
          Affymetrix HT Rat Focus Array 1372879_PM_at - -
          Affymetrix HT Rat Focus Array 1375766_at - -
          Affymetrix HT Rat Focus Array 1375117_at - -
          Affymetrix HT Rat Focus Array 1375766_PM_at - -
          Affymetrix HT Rat Focus Array 1375117_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100011432.rn.1 TC0100001467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100011434.rn.1 TC0100001467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100011433.rn.1 TC0100001467.rn.1 RTA TC
          Rat Expression Set 230 1375766_at - -
          Rat Expression Set 230 1375117_PM_at - -
          Rat Expression Set 230 1375766_PM_at - -
          Rat Expression Set 230 1372879_at - -
          Rat Expression Set 230 1375117_at - -
          Rat Expression Set 230 1372879_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706644 - -
          Rat Genome U34 Set rc_AI230166_at - -
          Rat Genome U34 Set rc_AI102030_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21871221 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348998 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0