Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455736_m1
          See other AASS GE Assays ›
          Gene Symbol:
          Aass
          Gene Name:
          aminoadipate-semialdehyde synthase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 50152005 - 50208935 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455736_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aass
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100963.1 NP_001094433.1 23 - 24 2713 59
          GenBank mRNA BC128771.1 - 23 - 24 2713 59

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          L-lysine catabolic process
          L-lysine catabolic process to acetyl-CoA via saccharopine
          molecular_function
          saccharopine dehydrogenase (NADP+, L-lysine-forming) activity
          saccharopine dehydrogenase (NAD+, L-glutamate-forming) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P347149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18531 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132233 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P347149 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P347149 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P347149 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P347149 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40579 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2660 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10861247 - -
          Affymetrix Rat Gene 1.0 ST Array 10861252 - -
          Affymetrix Rat Gene 1.0 ST Array 10861251 - -
          Affymetrix Rat Gene 1.0 ST Array 10861244 - -
          Affymetrix Rat Gene 1.0 ST Array 10861259 - -
          Affymetrix Rat Gene 1.0 ST Array 10861260 - -
          Affymetrix Rat Gene 1.0 ST Array 10861253 - -
          Affymetrix Rat Gene 1.0 ST Array 10861245 - -
          Affymetrix Rat Gene 1.0 ST Array 10861249 - -
          Affymetrix Rat Gene 1.0 ST Array 10861250 - -
          Affymetrix Rat Gene 1.0 ST Array 10861248 - -
          Affymetrix Rat Gene 1.0 ST Array 10861243 - -
          Affymetrix Rat Gene 1.0 ST Array 10861257 - -
          Affymetrix Rat Gene 1.0 ST Array 10861261 - -
          Affymetrix Rat Gene 1.0 ST Array 10861255 - -
          Affymetrix Rat Gene 1.0 ST Array 10861256 - -
          Affymetrix Rat Gene 1.0 ST Array 10861262 - -
          Affymetrix Rat Gene 1.0 ST Array 10861246 - -
          Affymetrix Rat Gene 1.0 ST Array 10861254 - -
          Affymetrix Rat Gene 1.0 ST Array 10861258 - -
          Affymetrix Rat Gene 1.1 ST Array 10861242 - -
          Rat Genome 230 2.0 Array 1377375_at - -
          Rat Genome 230 2.0 Array 1377375_PM_at - -
          Affymetrix HT Rat Focus Array 1377375_at - -
          Affymetrix HT Rat Focus Array 1377375_PM_at - -
          Rat Expression Set 230 1377375_at - -
          Rat Expression Set 230 1377375_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10861242 - -
          Rat Genome U34 Set rc_AA944898_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20772666 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0