Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01456693_g1
          See other RAB34 GE Assays ›
          Gene Symbol:
          Rab34
          Gene Name:
          RAB34, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 65448111 - 65452178 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01456693_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012140.1 NP_001012140.1 4 - 5 483 70
          GenBank mRNA BC086400.1 - 4 - 5 483 70
          FQ212591.1 - 5 - 6 533 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001153.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab34

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          endocytosis
          small GTPase mediated signal transduction
          metabolic process
          protein transport
          antigen processing and presentation
          lysosome localization
          Rab protein signal transduction
          Golgi to plasma membrane protein transport
          macropinocytosis
          protein localization to plasma membrane
          phagosome maturation
          phagosome-lysosome fusion
          GTPase activity
          GTP binding
          guanyl nucleotide binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120307 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120307 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P141288 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9498 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745211 - -
          Affymetrix Rat Gene 1.0 ST Array 10745207 - -
          Affymetrix Rat Gene 1.0 ST Array 10745203 - -
          Affymetrix Rat Gene 1.0 ST Array 10745204 - -
          Affymetrix Rat Gene 1.0 ST Array 10745209 - -
          Affymetrix Rat Gene 1.0 ST Array 10745205 - -
          Affymetrix Rat Gene 1.0 ST Array 10745206 - -
          Affymetrix Rat Gene 1.0 ST Array 10745208 - -
          Affymetrix Rat Gene 1.0 ST Array 10745210 - -
          Affymetrix Rat Gene 1.0 ST Array 10745212 - -
          Affymetrix Rat Gene 1.1 ST Array 10745202 - -
          Rat Genome 230 2.0 Array 1375558_at - -
          Rat Genome 230 2.0 Array 1376573_PM_at - -
          Rat Genome 230 2.0 Array 1375558_PM_at - -
          Rat Genome 230 2.0 Array 1376573_at - -
          Affymetrix HT Rat Focus Array 1375558_at - -
          Affymetrix HT Rat Focus Array 1376573_at - -
          Affymetrix HT Rat Focus Array 1375558_PM_at - -
          Affymetrix HT Rat Focus Array 1376573_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000010418.rn.1 TC1000001153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000010421.rn.1 TC1000001153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000010420.rn.1 TC1000001153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000010419.rn.1 TC1000001153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000010425.rn.1 TC1000001153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000010423.rn.1 TC1000001153.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000010426.rn.1 TC1000001153.rn.1 RTA TC
          Rat Expression Set 230 1375558_at - -
          Rat Expression Set 230 1375558_PM_at - -
          Rat Expression Set 230 1376573_at - -
          Rat Expression Set 230 1376573_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745202 - -
          Rat Genome U34 Set rc_AA924181_at - -
          Rat Genome U34 Set rc_AI178161_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20922946 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0