Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01458090_g1
          See other LOC100910336 GE Assays ›
          Gene Symbol:
          LOC100910336 Rps19
          Gene Name:
          40S ribosomal protein S19-like
          ribosomal protein S19
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.9: 16802280 - 16808037 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 81750928 - 81756629 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01458090_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100910336
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750647.3 XP_003750695.1 1 - 2 112 82
          Gene Symbol: Rps19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037346.2 NP_001032423.1 1 - 2 57 82
          GenBank mRNA CB720565.1 - 1 - 2 57 82

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001037346.2, XM_003750647.3 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100910336
          Click here to find siRNAs for Gene Rps19

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          monocyte chemotaxis
          rRNA processing
          translation
          nucleolus organization
          biological_process
          erythrocyte differentiation
          maturation of SSU-rRNA
          ribosomal small subunit biogenesis
          protein tetramerization
          positive regulation of respiratory burst involved in inflammatory response
          negative regulation of respiratory burst involved in inflammatory response
          RNA binding
          structural constituent of ribosome
          protein binding
          fibroblast growth factor binding
          protein kinase binding
          translation initiation factor binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P076880 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003994 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P107623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165964 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076880 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076880 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003994 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P107623 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076880 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P651334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99690 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892938 - -
          Affymetrix Rat Gene 1.0 ST Array 10705064 - -
          Affymetrix Rat Gene 1.0 ST Array 10705060 - -
          Affymetrix Rat Gene 1.0 ST Array 10705063 - -
          Affymetrix Rat Gene 1.0 ST Array 10778919 - -
          Affymetrix Rat Gene 1.0 ST Array 10904229 - -
          Affymetrix Rat Gene 1.0 ST Array 10799988 - -
          Affymetrix Rat Gene 1.0 ST Array 10705059 - -
          Affymetrix Rat Gene 1.0 ST Array 10740380 - -
          Affymetrix Rat Gene 1.0 ST Array 10904228 - -
          Affymetrix Rat Gene 1.0 ST Array 10783354 - -
          Affymetrix Rat Gene 1.0 ST Array 10704412 - -
          Affymetrix Rat Gene 1.0 ST Array 10705062 - -
          Affymetrix Rat Gene 1.0 ST Array 10795547 - -
          Affymetrix Rat Gene 1.0 ST Array 10705061 - -
          Affymetrix Rat Gene 1.1 ST Array 10799987 - -
          Affymetrix Rat Gene 1.1 ST Array 10783353 - -
          Affymetrix Rat Gene 1.1 ST Array 10704411 - -
          Affymetrix Rat Gene 1.1 ST Array 10892937 - -
          Affymetrix Rat Gene 1.1 ST Array 10795546 - -
          Affymetrix Rat Gene 1.1 ST Array 10705058 - -
          Affymetrix Rat Gene 1.1 ST Array 10904227 - -
          Affymetrix Rat Gene 1.1 ST Array 10740379 - -
          Affymetrix Rat Gene 1.1 ST Array 10778918 - -
          Rat Genome 230 2.0 Array 1371377_PM_at - -
          Rat Genome 230 2.0 Array 1371377_at - -
          Affymetrix HT Rat Focus Array 1371377_PM_at - -
          Affymetrix HT Rat Focus Array 1371377_at - -
          Rat Expression Set 230 1371377_at - -
          Rat Expression Set 230 1371377_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778918 - -
          Affymetrix Rat Gene 1.0 ST Array 10705058 - -
          Affymetrix Rat Gene 1.0 ST Array 10795546 - -
          Affymetrix Rat Gene 1.0 ST Array 10783353 - -
          Affymetrix Rat Gene 1.0 ST Array 10892937 - -
          Rat Genome U34 Set X51707cds_s_at - -
          Rat Genome U34 Set rc_AI177042_i_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370264 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0