Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01459362_m1
          See other LOC102555341 GE Assays ›
          Gene Symbol:
          LOC102555341 Arntl2
          Gene Name:
          aryl hydrocarbon receptor nuclear translocator-like protein 2-like
          aryl hydrocarbon receptor nuclear translocator-like 2
          Gene Aliases:
          -
          Arnt4, Bmal2
          Chromosome Location:
          Chr.4: 181205757 - 181222088 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 181103774 - 181158415 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01459362_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC102555341
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763468.1 XP_008761690.1 726 70
          Gene Symbol: Arntl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133391.1 NP_596882.1 10 - 11 1215 70
          GenBank mRNA AF327071.1 - 10 - 11 1221 70
          AY014837.1 - 9 - 10 1119 70
          AY014838.1 - 9 - 10 1128 70
          JQ361086.1 - 10 - 11 1242 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002067.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          signal transduction
          circadian rhythm
          positive regulation of circadian rhythm
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          molecular_function
          DNA binding
          DNA-binding transcription factor activity
          protein dimerization activity
          E-box binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P389444 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389444 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389444 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P389444 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085971 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389444 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P259977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103551 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859680 - -
          Affymetrix Rat Gene 1.0 ST Array 10859672 - -
          Affymetrix Rat Gene 1.0 ST Array 10859673 - -
          Affymetrix Rat Gene 1.0 ST Array 10859676 - -
          Affymetrix Rat Gene 1.0 ST Array 10859681 - -
          Affymetrix Rat Gene 1.0 ST Array 10859677 - -
          Affymetrix Rat Gene 1.0 ST Array 10859678 - -
          Affymetrix Rat Gene 1.0 ST Array 10859675 - -
          Affymetrix Rat Gene 1.0 ST Array 10859679 - -
          Affymetrix Rat Gene 1.0 ST Array 10859674 - -
          Affymetrix Rat Gene 1.1 ST Array 10859671 - -
          Rat Genome 230 2.0 Array 1394782_at - -
          Rat Genome 230 2.0 Array 1394782_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400014838.rn.1 TC0400002067.rn.1 RTA TC
          Rat Expression Set 230 1394782_at - -
          Rat Expression Set 230 1394782_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859671 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21204969 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357077 - -
          RatRef-12 Expression BeadChip ILMN_63822 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0