Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460303_m1
          See other TRPV3 GE Assays ›
          Gene Symbol:
          Trpv3
          Gene Name:
          transient receptor potential cation channel, subfamily V, member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 59829358 - 59863764 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460303_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpv3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025757.2 NP_001020928.2 8 - 9 1143 83
          XM_008767859.1 XP_008766081.1 8 - 9 3038 83
          XM_008767860.1 XP_008766082.1 7 - 8 1177 83
          GenBank mRNA AY325813.1 - 8 - 9 1162 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001026.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpv3

          Gene Ontology Categories:

          Function(s) Process(es)

          response to temperature stimulus
          response to heat
          negative regulation of hair cycle
          positive regulation of calcium ion import
          cation transmembrane transport
          ion channel activity
          cation channel activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59443 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47305 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130189 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4823 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72797 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735706 - -
          Affymetrix Rat Gene 1.0 ST Array 10735711 - -
          Affymetrix Rat Gene 1.0 ST Array 10735714 - -
          Affymetrix Rat Gene 1.0 ST Array 10735703 - -
          Affymetrix Rat Gene 1.0 ST Array 10735715 - -
          Affymetrix Rat Gene 1.0 ST Array 10735717 - -
          Affymetrix Rat Gene 1.0 ST Array 10735712 - -
          Affymetrix Rat Gene 1.0 ST Array 10735702 - -
          Affymetrix Rat Gene 1.0 ST Array 10735708 - -
          Affymetrix Rat Gene 1.0 ST Array 10735704 - -
          Affymetrix Rat Gene 1.0 ST Array 10735707 - -
          Affymetrix Rat Gene 1.0 ST Array 10735700 - -
          Affymetrix Rat Gene 1.0 ST Array 10735710 - -
          Affymetrix Rat Gene 1.0 ST Array 10735701 - -
          Affymetrix Rat Gene 1.0 ST Array 10735713 - -
          Affymetrix Rat Gene 1.0 ST Array 10735705 - -
          Affymetrix Rat Gene 1.0 ST Array 10735716 - -
          Affymetrix Rat Gene 1.1 ST Array 10735699 - -
          Affymetrix Rat Clariom S Assay PSR1000009403.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009402.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009398.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009397.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009406.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009400.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009399.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009405.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009401.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009404.rn.1 TC1000001026.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10735699 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358056 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0